Would you like to log in again?
Proto-Board Portable Workstation
Supplier: GLOBAL SPECIALTIES SE
PB-503C
Functionality and Performance of a Full Test Bench in a Compact Package
Ideal for analog, digital and microprocessor circuits, these proto-board workstations contain multiple features in one instrument, saving money spent on individual units and reducing space required in your lab. These complete electronics workstations are particularly helpful for design engineers and advanced technology applications in laboratories, test benches, and classrooms. Instrumentation, breadboarding, and a rugged DC regulated triple power supply are all integrated into a comprehensive workstation. The instrumentation features eight new individual selectable logic switches (offering settings of +5volts / +V and ground) with red (high) and green (low) buffered logic LED indicators. A 100KHz function generator with continuously variable sine, square, and triangle waveforms plus TTL pulse is also included. Instrumentation is additional equipped with two digital pulsers, audio experimentation speaker, SPDT switches, potentiometers, and debounced pushbutton switches. The breadboarding area has a total of 2520 uncommitted tie points, enough space for circuits containing 24 IC's of 14 pins or the equivalent. And, all the breadboarding sockets have an unlimited lifetime guarantee. Breadboard area is also removable with optional additional breadboard socket plates. The power supply offers three DC regulated supplies, one fixed at +5VDC and two variable from ±1.3 to ±15VDC. All three supplies are short-circuit protected, automatically current limiting the output to a safe level. The Portable Workstation is a complete electronics workstation housed in a rugged carrying case, making it ideal for students or engineers who need to transport materials from lab to project work area. The case folds into the size of a large briefcase, with convenient storage facilities for manuals, an optional multimeter, and the optional WK-1 wire jumper kit.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Proto-Board Workstations, Portable Workstation PB-503C 470121-170 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty