Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Select™ Pass-Thru Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific

Supplier: NOR-LAKE SCIENTIFIC
Select™
NSSP242WWF/4 NSSP242WWF/5 NSSP806WWW/0 NSSP242WWG/4 CASTERS NSSP242WWG/5 NSSP242WWF/0 STACKKIT NSSP242WWG/0 LEGS NSSP806WWG/5 NSSP806WWG/4 NSSP806WWF/5 SEISMICKIT TRIMKITC806SS RHD NSSP806WWG/0 TRIMKITL806 NSSP806WWF/4 TRIMKITL242SS NSSP524WWW/5 NSSP806WWF/0 NSSP524WWW/4 TRIMKITC524SS GLDOOR NSSP524WWG/5 TRIMKITL524 NSSP524WWF/5 NSSP524WWG/4 NSSP524WWF/4 TRIMKITL524SS NSSP524WWG/0 NSSP524WWF/0 TRIMKITC242SS TRIMKITL242 NSSP242WWW/4 TRIMKITC242 NSSP242WWW/5 LEVELERS NSSP242WWW/0 TRIMKITL806SS TRIMKITC524 TRIMKITC806 NSSP806WWW/4 NSSP806WWW/5 SDDOOR
Refrigerators are equipped with a digital 4 x 20 character LCD display microprocessor temperature controller, with audible and visual Hi/Lo temperature alarm

Glass door(s) on applicable models are full size and double pane. The glass door(s) are standard on all WWF suffix models and WWG suffix models (controller side). Solid door(s) on applicable models are full size. Solid door(s) are standard on WWW suffix models and WWG suffix models (noncontroller side). All 115 V units are supplied with a 5 – 15 P NEMA plug and 5 – 15 R NEMA receptacle. The 208/230 V units are supplied with a 6 – 15 P NEMA plug and 6 – 15 R NEMA receptacle. The 230 V units feature a Power Inlet Module (IEC60320) and require a locally supplied cord.

The temperature control range is adjustable from 2 to 10 °C (36 to 50 °F). Units are factory preset at 4 °C (39 °F). Safety features include, remote alarm contacts, continuous product temperature display, three sensor system (2 product,1 air), power supply switch, door ajar alarm, and 2 – 10 volt DC output for product temperature (selectable to air temperature). Password protection enables set-points and factory settings, with 100-event alarm logging. Integrated Hi/Lo product temperature alarm test, digital calibration for air and product, real time clock with date and time display, and a door opening counter with 24 hour automatic reset are also included. Adjustable alarm allows for beep or constant on (key pad selectable). Alarm volume is adjustable as well. Integraged monitor systems include, fan motor run-time monitor, defrost time monitor, and condenser/compressor run-time monitor. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner and door panel are painted white. Cabinet and solid doors are foamed-in-place with CFC-free, high-density polyurethane foam insulation. These heavy-duty doors feature pivot hinges, pull-door handles, magnetic door gaskets, and centered key door lock(s) (2 keys included). Additional standard features include, four casters (2 locking) on two- and four-door models, six casters (3 locking) on six-door models, three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, and top-mounted forced air refrigeration system. Automatic defrost and condensate evaporation. A 2.5 cm (1") diameter sensor access port is also included.

Certifications: UL and C-UL listed.

Ordering information: Refrigerators are supplied with 24-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).




 

ORDER

Expand All / Collapse All
Description Electrical Volume Dimensions Door Type Weight Supplier No. VWR Catalog Number Unit Price Quantity
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 725 L (25.6 cu. ft.) 69.8W x 88.5D x 202.2H cm (271/2 x 347/8 x 795/8") All Glass Doors 170 kg (375 lbs.) NSSP242WWF/0 89368-348 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 208/230V, 60Hz, 1PH 725 L (25.6 cu. ft.) 69.8W x 88.5D x 202.2H cm (271/2 x 347/8 x 795/8") All Glass Doors 170 kg (375 lbs.) NSSP242WWF/4 89368-420 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 230V, 50Hz, 1PH 725 L (25.6 cu. ft.) 69.8W x 88.5D x 202.2H cm (271/2 x 347/8 x 795/8") All Glass Doors 170 kg (375 lbs.) NSSP242WWF/5 89368-442 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 1577 L (55.7 cu. ft.) 140W x 88.5D x 202.2H cm (55 x 347/8 x 795/8") All Glass Doors 268 kg (590 lbs.) NSSP524WWF/0 89368-350 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 208/230V, 60Hz, 1PH 1577 L (55.7 cu. ft.) 140W x 88.5D x 202.2H cm (55 x 347/8 x 795/8") All Glass Doors 268 kg (590 lbs.) NSSP524WWF/4 89368-422 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 230V, 50Hz, 1PH 1577 L (55.7 cu. ft.) 140W x 88.5D x 202.2H cm (55 x 347/8 x 795/8") All Glass Doors 268 kg (590 lbs.) NSSP524WWF/5 89368-444 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 2430 L (85.8 cu. ft.) 209.5W x 91.1D x 202.2H cm (821/2 x 357/8 x 795/8") All Glass Doors 365 kg (805 lbs.) NSSP806WWF/0 89368-352 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 208/230V, 60Hz, 1PH 2430 L (85.8 cu. ft.) 209.5W x 91.1D x 202.2H cm (821/2 x 357/8 x 795/8") All Glass Doors 365 kg (805 lbs.) NSSP806WWF/4 89368-424 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 230V, 50Hz, 1PH 2430 L (85.8 cu. ft.) 209.5W x 91.1D x 202.2H cm (821/2 x 357/8 x 795/8") All Glass Doors 365 kg (805 lbs.) NSSP806WWF/5 89368-446 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 725 L (25.6 cu. ft.) 69.8W x 88.5D x 202.2H cm (271/2 x 347/8 x 795/8") Glass/Solid Doors 170 kg (375 lbs.) NSSP242WWG/0 89368-340 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 1577 L (55.7 cu. ft.) 140W x 88.5D x 202.2H cm (55 x 347/8 x 795/8") Glass/Solid Doors 268 kg (590 lbs.) NSSP524WWG/0 89368-342 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 2430 L (85.8 cu. ft.) 209.5W x 91.1D x 202.2H cm (821/2 x 357/8 x 795/8") Glass/Solid Doors 365 kg (805 lbs.) NSSP806WWG/0 89368-344 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 725 L (25.6 cu. ft.) 69.8W x 88.5D x 202.2H cm (271/2 x 347/8 x 795/8") All Solid Doors 170 kg (375 lbs.) NSSP242WWW/0 89368-356 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Select™ Pass-Thru Laboratory and Pharmacy Refrigerators 115V, 60Hz, 1PH 2430 L (85.8 cu. ft.) 209.5W x 91.1D x 202.2H cm (821/2 x 357/8 x 795/8") All Solid Doors 365 kg (805 lbs.) NSSP806WWW/0 89368-360 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed

Stacking Kits and Trim Kits
Description Supplier No. VWR Catalog Number Unit Price Quantity
Stacking Kits for Use with Models LF041WWW/0 and LR061WWW/0 STACKKIT 89401-076 Each Retrieving
Build-In Trim Kit for NSPT242, Per Door Side, Painted with Casters TRIMKITC242 89401-078 Each Retrieving
Build-In Trim Kit for NSPT242, Per Door Side, Painted with Casters TRIMKITC242SS 89401-080 Each Retrieving
Build-In Trim Kit for NSPT242, Per Door Side, Stainless Steel with Casters TRIMKITL242 89401-082 Each Retrieving
Build-In Trim Kit for NSPT242, Per Door Side, Painted with Legs TRIMKITL242SS 89401-084 Each Retrieving
Build-In Trim Kit for NSPT524, Per Door Side, Painted with Casters TRIMKITC524 89401-086 Each Retrieving
Build-In Trim Kit for NSPT524, Per Door Side, Self Standing with Casters TRIMKITC524SS 89401-088 Each Retrieving
Build-In Trim Kit for NSPT524, Per Door Side, Painted with Legs TRIMKITL524 89401-090 Each Retrieving
Build-In Trim Kit for NSPT524, Per Door Side, Self Standing with Legs TRIMKITL524SS 89401-092 Each Retrieving
Build-In Trim Kit for NSPT806, Per Door Side, Painted with Casters TRIMKITC806 89401-094 Each Retrieving
Build-In Trim Kit for NSPT806, Per Door Side, Self Standing with Casters TRIMKITC806SS 89401-096 Each Retrieving
Build-In Trim Kit for NSPT806, Per Door Side, Painted with Legs TRIMKITL806 89401-098 Each Retrieving
Build-In Trim Kit for NSPT806, Per Door Side, Self Standing with Legs TRIMKITL806SS 89401-100 Each Retrieving
Seismic Restraint Kit for Upright Cabinets, Requires Field Installation (2 Kits Required for 3-Door Cabinets) SEISMICKIT 89401-102 Each Retrieving

Doors, Casters and Legs
Description Supplier No. VWR Catalog Number Unit Price Quantity
Glass Door, Factory Installed (Applies Only to Upright Refrigerators) GLDOOR 89401-044 Each Retrieving
Solid Door, Factory Installed (Not Applicable to Sliding Door Models) SDDOOR 89401-046 Each Retrieving
Reverse Hinge Door RHD 89401-048 Each Retrieving
Legs in Lieu of Casters LEGS 89401-050 Each Retrieving
Casters in Lieu of Legs or Levelers (For Electrical Code Compliance Cord, Plug Should be Used with Casters) CASTERS 89401-052 Each Retrieving
Four Levelers in Lieu of Legs, 2", (Applicable to Undercounters) LEVELERS 89401-054 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is safer to human and environmental health

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
2" Diameter Access Port with Sleeve and Cover, Factory Installed (Not Applicable on Freezer or Flammable Storage Models) SLEEVE 89401-056 Each Retrieving
Secure Guard II Lock Assembly, Factory Installed for Prox Key/Key Pad (General-Purpose Undercounter LR & LF Series Only) PROXKEYGP 89401-108 Each Retrieving
Temperature Chart Recorder, 6", One Pen, Factory Installed (Standard on NSBR's & NSBF's) REC6P1 89400-988 Each Retrieving
Water Filter with Bracket and Installation Manual, Field Installation Required WATERFILTER 89401-104 Each Retrieving
1" Diameter Access Sensor Port with Plugs for NS Series Freezers and Refrigerators, Factory Installed (Not Applicable to LF, LR, LRF, or Flammable Refrigerators/Freezers) PORT 89401-058 Each Retrieving
Secure Guard II Lock Assembly, Factory Installed for Mag Key/Key Pad (General-Purpose Undercounter LR & LF Series Only) MAGKEYGP 89401-110 Each Retrieving
Temperature Chart Recorder, 6", Two Pens, Factory Installed (NSRI Chambers with Humidity) REC6P2 89400-990 Each Retrieving
Water Filter Cartridge, Siemens Universal II, IWT Ion Exchange Cartridge FILTER 89401-106 Each Retrieving
Pressure Relief Valve, Factory Installed (Not Applicable to Refrigerators or Flammable Storage Models) PRVALVE 89401-060 Each Retrieving
Secure Guard II Lock Assembly, Factory Installed for Prox Key/Key Pad, Select, Premier and NS Standard Models (Per Door) PROXKEY 89401-112 Each Retrieving
Temperature Chart Recorder, 6", One Pen, Free Standing REC6DP1 89400-992 Each Retrieving
Live Duplex Outlet with Vapor Proof Cover Factory Installed, 15A (Not Applicable to Flammable Storage or Undercounters) DUPLEX 89400-994 Each Retrieving
Secure Guard II Lock Assembly, Factory Installed for Mag Key/Key Pad, Select, Premier, and NS Standard Models (Per Door) MAGKEY 89401-114 Each Retrieving
White Interior INTWHITE 89401-062 Each Retrieving
4–20Ma Output for Freezers, Factory Installed, Standard Digital Display MA420F 89400-996 Each Retrieving
Secure Guard II Lock Assembly, Field Installed for Prox Key/Key Pad (General-Purpose Undercounter LR & LF Series Only) PROXKEYGPKIT 89401-116 Each Retrieving
4–20MA Output for Refrigerators, Factory Installed for Refrigerators with Standard Digital Display MA420R 89400-998 Each Retrieving
Secure Guard II Lock Assembly Field, Installed for Mag Key/Key Pad (General-Purpose Undercounter LR & LF Series Only) MAGKEYGPKIT 89401-118 Each Retrieving
4–20Ma Temperature Output, Factory Installed for SELECT, NSBR's, NSBF's, NSRI's and NSHI'S (Not Applicable to Undercounters) MA420PCO 89401-000 Each Retrieving
Enzyme Storage Bin, 23 x 4 x 4" BIN 89401-068 Each Retrieving
Secure Guard II Lock Assembly Field, Installed for Prox Key/Key Pad, Select, Premier, and NS Standard Models (Per Door) PROXKEYKIT 89401-120 Each Retrieving
Humidity Transfer Output, 0–10 Vapor Compression, Factory Installed for NSRI's VDC210PCO 89401-002 Each Retrieving
Secure Guard II Lock Assembly Field, Install for Mag Key/Key Pad, Select, Premier, and NS Standard Models (Per Door) MAGKEYKIT 89401-122 Each Retrieving
Export Crating Chipboard, Placed on All 4 Sides and on Top of Pallet EXPCRATE 89401-072 Each Retrieving
RS-485 Digital Communcation Port, Factory Installed for SELECT, NSBR's, NSBF's, NSRI's and NSHI's (Not Applicable to Undercounters) RS485PCO 89401-004 Each Retrieving
Secure Guard II Lock Software SECURESOFT 89401-124 Each Retrieving
Humidity Pan for HSHI Series Incubators PANHUMID 89401-074 Each Retrieving
RS-485 Digital Communcation Port, Factory Installed for Premier Series with XR70 Dixell Control (Not Applicable to Undercounters) RS485DIX 89401-006 Each Retrieving
Water Drip Pan for Right Hand Door Swing Models, Factory Installed DRIPPANRH331 89401-126 Each Retrieving
Hi/Lo Alarm with Remote Contacts and Buzzer, 4–20MA, Factory Installed for Freezers with Standard Digital Display ALARMF 89401-008 Each Retrieving
Water Drip Pan for Left Hand Door Swing Models, Factory Installed DRIPPANLH331 89401-128 Each Retrieving
Hi/Lo Alarm with Remote Contacts and Buzzer, Factory Installed for Refrigerators with Standard Digital Display ALARMR 89401-010 Each Retrieving
Water Drip Pan for Left Hand Door Swing Models, Factory Installed DRIPPANLH 89401-130 Each Retrieving
Hi/Lo Alarm with Remote Contacts and Buzzer, Factory Installed for Refrigerators & Freezers ALARMRF 89401-012 Each Retrieving
Water Drip Pan for Right Hand Door Swing Models, Factory Installed DRIPPANRH 89401-132 Each Retrieving
485 Communication Port, Factory Installed for Premier, NSPR, NSPF Series with Dixell XR70 Controller RS485DXL 89401-014 Each Retrieving
Remote Contacts, 4–20Ma, Factory Installed for Freezers with Standard Digitial Display (NSPF Premier Series) REMCONTF 89401-016 Each Retrieving
Thermometer Monitor with Digital Display and Alarm (Field Installation), Not Applicable to Flammable Storage Refrigerators or Freezers THERMALARM 89401-018 Each Retrieving
RH Display for NSHI Series Incubators RHDISPLAY 89401-020 Each Retrieving
Ethernet 10baseT (HTTP) Control/Monitoring Web Card ENET 89401-022 Each Retrieving
CO₂ Injection and Control For Lighted Chambers, Factory Installed Only INJECTCO2 89401-024 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES

Expand All / Collapse All

VWR® Customer On-site Calibration, Validation and PM Services
Description Details Includes Service Coverage Out-of-Service Coverage Supplier No. VWR Catalog Number Unit Price Quantity
VWR® NIST On-Site Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. Calibration at a single point using NIST traceable standards. Upon completion, a calibration form will be provided with “as found” and “as left” measurements. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-880 Each Retrieving
VWR® NIST On-Site PM and Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. PM and Calibration service includes cleaning of seals, fans, and gaskets. The chamber is inspected interiorly and exteriorly. After PM, technicians will calibrate the chamber as described in Calibration section. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-888 Each Retrieving
VWR® NIST Traceable Validate (IOQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. Validation (IQ/OQ) includes a standardized protocol procedure, calibration, tuning, IQ (verify design spec, system SOP, environmental conditions etc), and OQ (Perform empty chamber temperature mapping, open door and power loss testing). (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-950 Each Retrieving
VWR® NIST Traceable Validate (IOPQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. IQ/OQ will be performed as described in the (IQ/OQ) section. The PQ consists of loaded chamber temperature mapping for an additional 24 hours. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-942 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty