The Ohaus® Defender™ 7000 Series is an advanced multifunctional line of indicators with a rectangular base and bench scales. They are ideal for shipping and receiving, production, packing and general commercial, industrial, and lab applications. Combining an IP66 stainless steel T71XW indicator with all-stainless steel bases and columns, the Defender™ 7000 offers multi-functionality and multiple weighing units, alpha-numeric keypad, and enhanced software modes. With rugged construction, comprehensive yet intuitive software, multiple connectivity, battery operation, and configurable mounting, the Defender™ 7000 is a flexible one-stop solution for demanding weighing operations.
Component construction includes, IP66 stainless steel Indicator, 304 grade stainless steel platform pan with stainless steel form-and weld frame, IP67 stainless steel load cell, and stainless steel column
All units feature an indicator with hinged front cover for easy servicing, an alpha numeric keypad, and a platform with 5 or 6 point overload stops for 150% safe overload protection. Scales are also equipped with an LCD display with white LED backlight, alphanumeric line, multi-language options, and a three color checkweigh LED.
Weighing (6 Units + Custom Unit), Display Hold, Dynamic Weighing, Counting, Checkweighing, % Weighing, Auto-Tare, Accumulation, Library of up to 256 Records, and Optional Alibi Memory.
Wall bracket included.
Certifications: C-UL listed. NTEP Certificate of Conformance No. 09-102 and 99-203A3, Measurement Canada Notices of Approval AM-5762 and AM-5675.
Ordering information: Scales are supplied with internal universal power supply or optional internal NiMH rechargeable battery, and an RS232 Interface with GMP and Time-Date. Optional RS422/485, AC/DC Relay, and 2nd RS232 features are available.