The Ohaus® Defender™ 7000 Series is an advanced multifunctional line of indicators, bases, and bench scales that are ideal for shipping and receiving, production, packing and general commercial, industrial, and lab applications. Combining an IP66 stainless steel T71XW indicator with all-stainless steel bases and columns, the Defender™ 7000 offers multi-functionality and multiple weighing units. An alpha-numeric keypad and enhanced software modes further facilitate easy operation. With rugged construction, comprehensive yet intuitive software, multiple connectivity, battery operation, and configurable mounting, the Defender™ 7000 is a flexible one-stop solution for a variety of demanding weighing operations.
Component construction includes, IP66 stainless steel Indicator, 304 grade stainless steel platform pan with stainless steel form-and weld frame, IP67 stainless steel load cell, and stainless steel column
All units feature an indicator with hinged front cover for easy servicing, an alpha numeric keypad, and a platform with 5 or 6 point overload stops for 150% safe overload protection. Scales are also equipped with an LCD display with white LED backlight, alphanumeric line, multi-language options, and a three color checkweigh LED.
Weighing (6 Units + Custom Unit), Display Hold, Dynamic Weighing, Counting, Checkweighing, % Weighing, Auto-Tare, Accumulation, Library of up to 256 Records, and Optional Alibi Memory.
Wall bracket included.
Certifications: C-UL listed. NTEP Certificate of Conformance No. 09-102 and 01-048AL, Measurement Canada Notices of Approval AM-5762 and AM-5719.
Ordering information: Scales are supplied with internal universal power supply or optional internal NiMH rechargeable battery, and an RS232 Interface with GMP and Time-Date. Optional RS422/485, AC/DC Relay, and 2nd RS232 features are available.