The only scale in its class with touchless sensors allows for hands-free operation. The Navigator® series offers a unique combination of features, including touchless sensors, EC Type approved models, optional internal rechargeable battery, LED or LCD display, ultra-fast response time, and superior overload protection for use in a wide variety of weighing applications.
- Applications include weighing (14 units), percentage weighing, check weighing, parts counting, accumulation/totalization, and display hold
Navigator® scales feature dual multi-functional touch-free infrared sensors, front level indicator (lit during power up), non-slip adjustable feet, stability indicator, external menu lock switch, shipping transportation lock, integrated security loop, and superior overload protection
Additional standard features include, six full segment LCD white auto backlight or red LED, both with multiple icons and indicators. The 1 second stabilization time provides results in a snap.
All scales feature ABS plastic construction with a stainless steel pan.
Certifications: C-UL listed.
Ordering information: Units feature mains or battery operation with auto shut-off or optional internal rechargeable battery on NVL and NVT. Easy to install USB, RS232, and Ethernet interface options.