Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Navigator® XL Portable Balance, Ohaus®

Supplier: OHAUS SCALE CORP
Navigator®
NVL10001 NVL20000 NVL511 NVL1101 NVL2101 NVL5101
The only scale in its class with touchless sensors allows for hands-free operation. The Navigator® series offers a unique combination of features, including touchless sensors, EC Type approved models, optional internal rechargeable battery, LED or LCD display, ultra-fast response time, and superior overload protection for use in a wide variety of weighing applications.

  • Applications include weighing (14 units), percent weighing, check weighing, parts counting, accumulation/totalization, and display hold

Navigator® scales feature dual multi-functional touch-free infrared sensors, front level indicator (lit during power up), non-slip adjustable feet, stability indicator, external menu lock switch, shipping transportation lock, integrated security loop, and superior overload protection

Additional standard features include, six full segment LCD white auto backlight or Red LED, both with multiple icons and indicators. The 2 second stabilization time provides results in a snap.

All scales feature ABS plastic construction with a stainless steel pan.

Certifications: C-UL listed. NTEP and Measurement Canada approved models meet or exceed Class III accuracy requirements.

Ordering information: Units feature mains or battery operation with auto shut-off or optional internal rechargeable battery on NVL and NVT. Easy to install USB, RS232, and Ethernet interface options.



 

ORDER

Expand All / Collapse All
Weighing Capacity Pan/Platform Size Readability Supplier No. VWR Catalog Number Unit Price Quantity
510 g (1.1 lbs.) 194 x 203 mm (7.6 x 8") 0.1 g (0.0002 lbs.) 83033058 89203-160 Each Retrieving
cUL Listed
1100 g (2.4 lbs.) 194 x 203 mm (7.6 x 8") 0.1 g (0.0002 lbs.) 83033062 89203-162 Each Retrieving
cUL Listed
2100 g (4.6 lbs.) 194 x 203 mm (7.6 x 8") 0.1 g (0.0002 lbs.) 83033066 89203-164 Each Retrieving
cUL Listed
5100 g (11.2 lbs.) 194 x 203 mm (7.6 x 8") 0.5 g (0.001 lbs.) 83033070 89203-166 Each Retrieving
cUL Listed
10,000 g (22 lbs.) 194 x 203 mm (7.6 x 8") 1 g (0.002 lbs.) 83033074 89203-168 Each Retrieving
cUL Listed
20,000 g (44 lbs.) 194 x 203 mm (7.6 x 8") 1 g (0.002 lbs.) 83033078 89203-170 Each Retrieving
cUL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty