VX Series Washdown Floor Scales, Ohaus®
Supplier: OHAUS SCALE CORP
80253294 80253298 80253296 80253297 80253295
97051-866EA 3157.22 USD
97051-866 97051-874 97051-870 97051-872 97051-868
VX Series Washdown Floor Scales, Ohaus®
Balances and Scales
VX Series Washdown Floor Scales are equipped with durable T32XW Washdown Indicators
  • Weighing
  • Parts counting
All scales feature a durable, epoxy painted steel frame and platform with NEMA4X/IP65 stainless steel indicator, NEMA4X/IP67 sealed stainless steel junction box, and IP67 sealed loadcells.
Scales offer selectable weight display in kg or lb. measurements. All scales feature a counting mode, leveling bubble, and an RS232 output for printing or connection to a PC. The 2500L and 5000L models are NTEP certified.
Certifications: C-UL listed. NTEP, Measurement Canada Notice of Approval AM-5449.
Ordering information: Scales are supplied with an AC Power adapter and rechargeable battery. Units also include an RS232 output for printing or connection to a PC.