Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: Expedeon
GK-42012-10 MGP15-100RDN MGP15-100 GK-42020-10 MGP15-100DN GK-00812-10 MGE15-1010 GK-01220-10 MG15-0712M MG15-1520 MG15-0720 MGP15100RLFP MG15-1020 MGE15-0710 MG15-1512M MG15-1012M DCX8-MC4 MGE15-1510 MGP15-100R MGP15100LFP GK-00820-10 GK-01212-10




 

ORDER

Expand All / Collapse All
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Comb for 10 x 15 cm, 0.75 mm x 12 +2 Wells Tooth Width: 2.5 mm + 7 mm/marker, Well Volume: 45 µL + 15 µL/marker, No. of Teeth: 12 + 2 markers, Thickness: 0.75 mm Comb and 0.75 mm Spacer MG15-0712M CBMG15-0712M Each Retrieving
Comb for 10 x 15 cm, 0.75 mm x 20 Wells Tooth Width: 4 mm, Well Volume: 30 µL, No. of Teeth: 20, Thickness: 0.75 mm Comb and 0.75 mm Spacer MG15-0720 CBMG15-0720 Each Retrieving
Comb for 10 x 15 cm, 1.0 mm x 12 +2 Wells Tooth Width: 2.5 mm + 7 mm/marker, Well Volume: 60 µL + 20 µL/marker, No. of Teeth: 12 + 2 markers, Thickness: 1 mm Comb and 1 mm Spacer MG15-1012M CBMG15-1012M Each Retrieving
Comb for 10 x 15 cm, 1.0 mm x 20 Wells Tooth Width: 4 mm, Well Volume: 40 µL, No. of Teeth: 20, Thickness: 1 mm Comb and 1 mm Spacer MG15-1020 CBMG15-1020 Each Retrieving
Comb for 10 x 15 cm, 1.5 mm x 12 +2 Wells Tooth Width: 2.5 mm + 7 mm/marker, Well Volume: 90 µL + 30 µL/marker, No. of Teeth: 12 + 2 markers, Thickness: 1.5 mm Comb and 1.5 mm Spacer MG15-1512M CBMG15-1512M Each Retrieving
Comb for 10 x 15 cm, 1.5 mm x 20 Wells Tooth Width: 4 mm, Well Volume: 60 µL, No. of Teeth: 20, Thickness: 1.5 mm Comb and 1.5 mm Spacer MG15-1520 CBMG15-1520 Each Retrieving
Gel Wrap Gasket 0.75 mm for 10 x 15 cm Thickness: 0.75 mm Comb and 0.75 mm Spacer MGE15-0710 CBMGE15-0710 Each Retrieving
Gel Wrap Gasket 1.0 mm for 10 x 15 cm Thickness: 1 mm Comb and 1 mm Spacer MGE15-1010 CBMGE15-1010 Each Retrieving
Gel Wrap Gasket 1.5 mm for 10 x 15 cm Thickness: 1.5 mm Comb and 1.5 mm Spacer MGE15-1510 CBMGE15-1510 Each Retrieving
Glass Plate Set 10 x 15 cm Casting Chamber - MGP15-100 CBMGP15-100 Each Retrieving
Glass Plate Set 10 x 15 cm Casting Chamber Back Plate Borosilicate - MGP15-100DN CBMGP15-100DN Each Retrieving
Glass Plate Set 10 x 15 cm Casting Chamber Borosilicate - MGP15100LFP CBMGP15100LFP Each Retrieving
Glass Plate Set 10 x 15 cm Gel Wrap - MGP15-100R CBMGP15-100R Each Retrieving
Glass Plate Set 10 x 15 cm Gel Wrap Back Plate Borosilicate - MGP15-100RDN CBMGP15-100RDN Each Retrieving
Glass Plate Set 10 x 15 cm Gel Wrap Borosilicate - MGP15100RLFP CBMGP15100RLFP Each Retrieving
Multiple Gel Casting Chamber 4 place for 10 x 15 cm Gels - DCX8-MC4 CBDCX8-MC4 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty