Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Premier™ Sliding Glass Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific

Supplier: NOR-LAKE SCIENTIFIC
Premier™
NSPR502WWG/0 NSPR502WWG/5
Refrigerators are equipped with a digital LED display microprocessor and temperature controller with audible and visual Hi/Lo temperature alarm

The 115V unit is supplied with a 5-15P NEMA plug and 5-15R NEMA receptacle. The 230V unit features a Power Inlet (IEC60320) module and requires a locally supplied cord.

They offer an adjustable temperature control range from 2 to 10°C (36 to 50°F). All models are factory preset at 4°C (39°F). Safety features include, remote alarm contacts, continuous product temperature display, product sensor with glycerin bottle, and power supply switch. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner is painted white. Cabinet is foamed-in-place with CFC-free, high-density polyurethane foam insulation. The glass doors are full-size and double pane. These self-closing glass doors have a "hold open" feature, with centered key door lock(s) (2 keys included). Additional standard features include, four casters (2 locking), three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, and top-mounted forced air refrigeration system.

Certifications: All units are UL and C-UL listed.

Ordering information: Refrigerators are supplied with an 18-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).




 

ORDER

Expand All / Collapse All
Description Electrical Volume Dimensions No. of Doors Weight Supplier No. VWR Catalog Number Unit Price Quantity
Premier™ Sliding Glass Door Laboratory and Pharmacy Refrigerator 115V, 60Hz, 1PH 1380 L (48.7 cu. ft.) 140W x 86.4D x 202.2H cm (55 x 34 x 79⅝") 2 Doors 243 kg (535 lbs.) NSPR502WWG/0 89368-116 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Premier™ Sliding Glass Door Laboratory and Pharmacy Refrigerator 230V, 50Hz, 1PH 1380 L (48.7 cu. ft.) 140W x 86.4D x 202.2H cm (55 x 34 x 79⅝") 2 Doors 243 kg (535 lbs.) NSPR502WWG/5 89368-118 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is safer to human and environmental health

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty