Premier™ Solid Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Premier™
NSPR803WWW/5 NSPR522WWW/0 NSPR803WWW/0 NSPR331WWW/0 NSPR241WWW/0 NSPR522WWW/5 NSPR241WWW/5 NSPR331WWW/5
89368-176EA 13215.15 USD
89368-176 89368-170 89368-174 89368-276 89368-166 89368-172 89368-168 89368-278
Premier™ Solid Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Refrigerators are equipped with a digital LED display microprocessor and temperature controller with audible and visual Hi/Lo temperature alarm
All 115V units are supplied with a 5-15P NEMA plug and 5-15R NEMA receptacle. The 230V units feature a Power Inlet (IEC60320) module and require a locally supplied cord.
They offer adjustable temperature control from 2 to 10°C (36 to 50°F). All models are factory preset at 4°C (39°F). Safety features include, remote alarm contacts, continuous product temperature display, product sensor with glycerin bottle, and power supply switch. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner and interior door panel are painted white. Cabinet and solid door are foamed-in-place with CFC-free, high-density polyurethane foam insulation. The heavy-duty solid door(s) feature pivot hinges, full-length pull-door handle(s), magnetic door gaskets, and centered key door lock(s) (2 keys included). Additional standard features include, four casters (2 locking) on one- and two-door models, six casters (3 locking) on three-door models, three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, top-mounted forced air refrigeration system, automatic defrost, and condensate evaporation.
Certifications: All units are UL and C-UL listed.
Ordering information: Refrigerators are supplied with an 18-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).