Premier™ Glass Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Premier™
NSPR241WWG/5 NSPR241WWG/0 NSPR803WWG/5 NSPR331WWG/5 NSPR803WWG/0 NSPR522WWG/5 NSPR331WWG/0 NSPR522WWG/0
89368-180EA 7043.54 USD
89368-180 89368-178 89368-188 89368-274 89368-186 89368-184 89368-272 89368-182
Premier™ Glass Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Refrigerators are equipped with a digital LED display microprocessor and temperature controller with audible and visual Hi/Lo temperature alarm
All 115V units are supplied with a 5-15P NEMA plug and 5-15R NEMA receptacle. The 230V units feature a Power Inlet (IEC60320) module and require a locally supplied cord.
They offer adjustable temperature control from 2 to 10°C (36 to 50°F). All models are factory preset at 4°C (39°F). Safety features include, remote alarm contacts, continuous product temperature display, product sensor with glycerin bottle, and power supply switch. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner is painted white. Cabinet is foamed-in-place with CFC-free, high-density polyurethane foam insulation. The glass doors are full-size and double pane, with anticondensate and heater wires. These heavy-duty doors feature pivot hinges, pull-door handles, magnetic door gaskets, and centered key door lock(s) (2 keys included). Additional standard features include, four casters (2 locking) on one- and two-door models, six casters (3 locking) on three-door models, three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, top-mounted forced air refrigeration system, automatic defrost, and condensate evaporation.
Certifications: All units are UL and C-UL listed.
Ordering information: Refrigerators are supplied with an 18-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).