Cam Fitting, Stainless Steel, Male Cam×3/4" NPT(M)
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable stainless steel
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.