Masterflex® Submersible Dewatering Pump, 68 GPM/57 ft, 304 SS, Manual; 115 VAC
Supplier: Avantor Fluid Handling
Semi-open impeller design pumps solids up to 3/8" in slurries and up to 81 GPM.
- Stainless steel construction resists corrosion and rust
- Pumps with automatic operation have float switch; manual operation models activate with a switch
- Air-filled motors have permanent split capacitor design with built-in thermal protection and automatic reset
- Maximum flow rate 68 GPM
- Maximum feet of head 57 ft
- Operates on 115 VAC
Operate pumps either fully or partially submerged. Pumps are ideal for slurries and dirty or semi-dirty waters.
Delivery information: Includes 20 ft (6.1 m) water-resistant #16 AWG cord.