Masterflex® Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 100/PK
Supplier: Avantor Fluid Handling
Regulate flow without any leaking.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Three-way stopcock/flow diverter with male luer lock connections
  • HDPE handle rotates 180°
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.