MicroLiter Chromatography Supplies
MicroLiter Analytical Supplies, Inc., a WHEATON Brand, was founded in 1996 to supply autosampler vials and consumables for the chromatography autosampler market, including consumable vials, closures with septa, inserts, and microplates.  MicroLiter offers Certified Class 10,000 clean room packaged autosampler vials and inserts, ensuring minimal particulates from product packaging.  A comprehensive offering of crimp seals, screw thread and snap caps, with a variety of septa configurations, are packaged in tubes to allow the use of a bench top dispenser.  This unique dispenser minimizes environmental contamination to the closures once the packaging is opened.
Request a Sample
Product selection
MicroLiter Certified Clean Vials
MicroLiter Analytical Supplies, Inc., a WHEATON Brand, was founded in 1996 to supply autosampler vials and consumables. MicroLiter offers Certified Class 10,000 clean room packagedautosampler vials and inserts to ensure minimal particulates.
MicroLiter Component Kits
Component kits save time by providing all the necessary components at your fingertips. Component kits contain 100 vials, 100 caps and 100 septa.
MicroLiter Closures
A comprehensive offering of crimp seals, screw thread and snap caps, with a variety of septa configurations, are packaged in tubes to allow the use of a bench top dispenser.
MicroLiter Limited Volume Inserts
Limited volume inserts are ideal for applications where there is a small amount of sample. MicroLiter offers both glass inserts and ready to use microplates with glass inserts.
MicroLiter Plate Sampling System
The MicroLiter Plate Sampling System (MPSS) is a patented system of microplates with glass inserts and re-sealable closures. The MPSS allow easy sample prep and transfer of samples with minimal amounts of human interaction to the analytical instrument.
E-Z Decappers™ Hand-Operated Decappers
E-Z Decappers provide acomfortable way to remove seals quickly and easily. Decappers feature a cushioned ergonomic handle that helps reduce hand fatigue.
E-Z Crimper™ Hand-Operated Crimpers
E-Z Crimpers provide consistent sealing and a short, cushioned ergonomic handle that helps reduce hand fatigue and adds comfort. E-Z Crimpers are available for 8, 11, 13 and 20mm seal sizes.
Syringe Filters and Disposable Syringes
MicroLiter’s syringe filters are available in a variety of media types and pore sizes. Disposable syringes have polyethylene plungers for a wide range of chemical compatibility.