Bottles for every lab
Product selection
Media Bottles
WHEATON™ Media Bottles are a mainstay in laboratories across the world. All WHEATON Media Bottles with caps feature sloping shoulders to facilitate pouring and attached pour rings to eliminate dripping.
Serum Bottles
WHEATON Serum Bottles are offered in sizes that range from 2mL to 500mL. They are made from borosilicate glass that conforms to USP Type I requirements.
Plastic Bottles
Leak-resistant, HDPE, LDPE, PP and much more.
Duran Bottles
As one of the world‘s top borosilicate glass, Duran® glass has proven itself excptional in both laboratories as well as in industrial applications.