Would you like to log in again?
VWR Returnable Container Advantage Program (ReCAP)
VWR Returnable Container Advantage Program (ReCAP)
PERFORMANCE | CONVENIENCE | SAFETY
Built on the foundation of supply chain innovation, VWR’s Returnable Container Advantage Program (ReCAP) reliably delivers performance, convenience, and safety for your high-purity solvents.
The program starts with your choice of sealed 200L stainless steel containers, filled with BDH high-purity Acetonitrile or Methanol (choose your specification). Once at your facility, solvent can be safely dispensed without risking product contamination, spillage, or personal exposure. When the container is almost empty, let us know and we will replace it with a full one so your work remains uninterrupted.
Request More Information about the VWR ReCAP Program
The Advantage in VWR ReCAP
  • Inert gas pressurizes container to smoothly dispense
    high-purity solvent
  • High quality and safety with 304 stainless steel
  • Tamper-evident seal for peace of mind
  • Relief valve for pressure regulation
  • Convenience through Swagelok® Quick-Connects
  • Serialized containers for precise tracking
  • Integrated fork channels for hassle-free transport
  • Consumes 50% less storage space than the same volume in 4L bottles
  • Reduced effort and time for hazmat and packaging disposal
Exclusive Double-Wall Container Features
  • Innovative double-wall, stainless steel construction protects the inner vessel and provides secondary containment in the event of an inner leak
  • Insulation reduces pressure build-up during a fire by up to 90%
  • Split knuckle allows better access for inspection and cleaning
  • Multiple mobility options
  • Dual pressure-relief valves for additional safety