qScript® cDNA Synthesis Kits
Our advanced reverse transcriptase technology is synonymous with maximum yield and sensitivity. qScript first-strand cDNA synthesis reagents are rigorously optimized to provide sensitive and reliable detection of low-abundance RNA for qPCR assays. A broad linear dynamic range of input RNA provides reliable assay sensitivity for robust gene expression analysis.
Product selection
qScript cDNA SuperMix
High-performance qScript RT pre-blended with ribonuclease-inhibitor protein and optimized primer blend, ensuring integrity of low-abundance RNA and unbiased representation.
qScript XLT cDNA SuperMix
Next-gen qScript XLT with novel hot start RT technology in an easy-to-use 5x master mix for superior 2-step RT-qPCR assay performance and reproducibility.
qScript XLT 1-Step RT-qPCR ToughMix
A highly sensitive, ready-to-use master mix for quantitative reverse transcription PCR (RT-qPCR) of RNA templates using probe-based detection methods.
qScript One-Step SYBR Green RT-qPCR
Convenient and highly sensitive solution for quantitative RT-PCR of RNA templates (RT-qPCR) using SYBR Green I dye detection and gene-specific primers.