Would you like to log in again?
Chemicals & Flux
Chemicals & Flux
VWR is a manufacturer of laboratory chemicals and has a complete line of acids, solvents, salts and chemical solutions to suit the demanding requirements of any mining laboratory. Many of our laboratory chemicals are produced at Anachemia’s two chemical manufacturing plants in Lachine, Quebec, and in Rouses Point, New York. Anachemia chemicals are consistently rated as the most popular chemical brand sold by VWR to mining customers.
We also specialize in fire assay chemicals and custom made flux recipes using the highest purity litharge available. Our fluxes have gold levels as low as less than 1 ppb. Anachemia fluxes are recognized globally for their quality and consistency. Our fire assay fluxes come in a variety of package sizes and custom packaging.
Anachemia also has a long history of providing custom chemical solutions for customers around the world. Ask your sales representative about VWR/Anachemia custom chemical solutions.