Would you like to log in again?
Environmental Testing solutions for Air, Water, and Soil
Environmental Testing solutions for Air, Water, and Soil
The demand and need for Environmental Testing continues with increased regulations, push for sustainable natural resources and need to prevent environmental conditions which harm health of the population. Whether you are testing water, air, or soil, VWR is proud to bring you a strong portfolio of chromatography, chemicals, consumables and tools you can use everyday.
Contact Local Team
Environmental Analysis Made Easy with BDH COD Vials
VWR Chemicals-BDH introduces new Chemical Oxygen Demand (COD) Vials for Environmental Testing!
The vials contain premeasured digestion reagents suited to your expected COD range, so there is no measuring of harmful chemicals - simply add your sample to the vial and digest. Vials come in packs of 25 or 150, multiple ranges, and mercury-free versions depending on your specific reaction. 
Find the Vial Range for your Application