Laboratory Furniture FAQ
VWR offers standing and sitting height casework, 35.75" and 30" respectively for the metal VWR Contour casework. Other heights are available.
Note: dimensions for cabinet heights do not include worktop thickness.
18", 24", 30", 36", 42", 48", 52", and 60" widths. Other custom widths are available.
Yes, VWR offers ADA-compliant casework, along with ADA-compliant sinks, faucets, fume hoods, and more.
Standard colors are #61 Light Neutral and #78 Snow White. Other colors are available upon request.
No, worktops are ordered separately as there are many choices of work surfaces, colors, and sizes available to meet your specific needs.
VWR offers 24" and 31" depths, and 54" and 60" depths for islands and peninsulas. Standard cabinet depth is 22" with a 1" front overhang, which allows a 1" or 8" space at the rear of the cabinet for plumbing and/or electrical.
Although we highly recommend that you use one of our highly trained installer; please contact your VWR furniture specialist to request an installation manual.
VWR offers a variety of fume hoods to meet your requirements.
VWR offers a quick-turnaround Rediship Program. Contact your VWR Furniture Specialist for more information.