Go with the FLOW | Smart. Effective. Innovative.
Click to play Flowmi video
See Flowmi in action
Flow Cytometry Preparation
Scienceware® Flowmi™ Cell Strainers provides fast, efficient filtering of small volume samples (up to 1000µl) while providing reduced risk of cytometer clogs.
Key Features and Benefits:
  • Preserves volume when filtering small sample volumes
  • Removes cell aggregates and creates uniform single cell suspensions
  • Decreases the potential for clogging of FLOW or FACS* instruments by effectively filtering cellular debris
  • Sterile, 40µm or 70µm porosity
  • Fits a wide variety of 1000µl Pipette tips
Recommended for use with samples having a maximum concentration of 2 million cells/ml (40µm) or 4 million cells/ml (70µm)