Controlled Environments
Products including Tyvek® for use in critical environments within the biotechnology, pharmaceutical, microelectronics, semiconductor, food and beverage, medical device, and other industries.
Sterile Cleanroom Manufacturing
Tyvek® IsoClean® Garments and Tyvek® Micro-Clean® 2-1-2 Garments
  • Comfortable, lightweight and durable
  • Individually packaged garments offered as sterile or clean-processed & sterile
  • Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
  • Serged or bound seam styles are available
Learn more
Non-Sterile Cleanroom Manufacturing
Tyvek® IsoClean®, Tyvek® Micro-Clean® 2-1-2, ProClean® and ProShield® Garments
  • Delivers the particle barrier, durability and comfort in use
  • Available as individually packaged and clean processed as well as bulk packaged
  • Variety of fabrics to meet your specific applications and needs
Learn more
Controlled Environment and General Protection
Tyvek® IsoClean®, ProClean® and ProShield® Garments
  • Delivers the particle barrier, durability and comfort in use
  • Offers protection against non-hazardous particulates and light liquid splash
  • Lightweight and low linting
Learn more