Clear, sterile collection bottle pre-dosed with sodium thiosulfate
The Corning Gosselin water sample collection bottle is designed to meet water sample collection requirements of the United States EPA and Health Canada. The lightweight polyethylene terephthalate (PET) 250 mL bottle is pre-dosed with pentahydrated sodium thiosulfate, which provides enhanced stabilization of chlorinated water. The 160 mg/L dosage is in accordance with the 22nd edition of the Standard Method published by the American Public Health Association (APHA), the American Water Works Association (AWWA), and the Water Environment Federation (WEF). The bottle will neutralize up to 15 mg/L of free chlorine in water samples.
Key Features
  • Sterile
  • PET is transparent and break resistant
  • Molded graduations for accurate volume sample collection
  • Powdered sodium thiosulfate is easier to dissolve than tablets
  • Labeled with lot number, expiration date, and sodium thiosulfate dosage information
  • Convenient shrink-wrapped packs of 24 bottles
  • Wide neck for easier sample collection
  • Easy grip cap
  • Tamper-evident screw cap to ensure integrity until collection
Order Here
Corning® Gosselin™ Round HDPE Storage Bottles
High density polyethylene bottles with wide neck
  • HDPE storage bottles are easier to handle and are ideal for sampling and storage.
  • HDPE resin is lightweight and break-resistant
  • Bottles are leakproof
Key features:
  • Good for storage of liquid, semi-liquid, and solid forms
  • Available in six sizes: 50 mL, 100 mL, 150 mL, 250 mL, 500 mL, and 1L
  • Large neck opening provides easier access for liquid transfer
  • Sterile
  • Bottles are food contact compliant
Order
Microbiology Solutions
From sampling and preparation to testing and analysis, our technical expeerts understand your challenges and the changing regulatory requirements that shape your day-to-day processes.
Sampling, Preparation, Testing, Analysis, and Disposal
Related Information