Would you like to log in again?
Cole-Parmer® Masterflex® tubing and pumps
Improve fluid conditions and productivity in your lab with a Masterflex solution. From low-cost, fixed-speed pumps to advanced-controlled models suitable for critical metering and dispensing applications, Masterflex has the answer to your needs. VWR offers a wide selection of single or multi-channel Masterflex peristaltic pumps, as well as the most suitable options in tubing to be used with peristaltic pumps.
Manufactured for high-purity systems, peristaltic pumps are designed for fluids to have a non-contaminated flow path for the transfer of sensitive cell cultures and hazardous or heavy-duty liquids. With only a few moving parts they are easy to maintain, can be installed in minutes, and to pump a different fluid you only need to change the tubing.
Here are 3 reasons why to choose a Masterflex peristaltic pump:
  • A contamination-free system for pumping fluids, even those with high viscosities
  • Low maintenance, self-priming, and non-siphoning
  • Gentle, low-shear pumping operation ideal for handling live cells, large proteins, suspensions, and other shear-sensitive fluids