Would you like to log in again?
VWR Electronic Pipette…Pipetting Made Simple
VWR Electronic Pipette…Pipetting Made Simple
Lightweight ergonomics, easy to use, precise and reliable. All of the enhanced features you look for in an electronic pipettor with the familiar look and feel of a mechanical pipette.
  • Ease-of-use: Only 2 buttons and 5 pipetting modes including the CUSTOM Mode which allows to create pipetting protocols easily and rapidly
  • Advanced Ergonomics: Lightweight and Perfectly Balanced, it requires virtually no force to aspirate and dispense samples
  • Fully Motorized Piston Drive: For an even and regular sample delivery across each channel
  • VWR Two-Year Warranty
Contact your VWR sales representative today and see what we mean when we say…Pipetting Made Simple.
Request A Product Demonstration
VWR Electronic Pipette Instruction Video
Watch Now >
VWR Electronic Pipette More Details and Ordering Information
Have you already purchased your new VWR Electronic Pipette?
Congratulations on your purchase, we are certain you will be pleased!
Download User Manual
Download your FREE VWR Protocol Builder Software