Your session will expire in 15 minutes (at 08:20 PM).
Would you like to keep your session alive?
Would you like to log in again?
Bottle, GL Thread, Laboratory, Safety Coated, Ace Glass Incorporated
Supplier: Ace Glass
https://us.vwr.com/stibo/web/std.lang.all/07/37/8740737.jpg 100ML PLAS COATED LAB BOTTLE
Click to enlarge
These graduated laboratory bottles are chemically resistant and stable. Supplied with or without red PTFE-lined PBT cap and a removable pouring ring that allows for drip-free operations. Safety coated for protection against mechanical damage and to prevent leakage of contents should the glass break. For use up to 135°C. Not suitable for vacuum or pressure use. Can be hot air sterilized or autoclaved. Fabricated from borosilicate glass.
Log In to see your contract pricing and availablity
ORDER
Expand All / Collapse All
Outer Diameter GL Thread Size Capacity Description Supplier No. Height with Cap VWR Catalog Number Unit Price Quantity
56 mm 45 100 mL Laboratory GL Thread Bottle, With Cap, Safety Coated 5539-105 108 mm 89051-784 Case of 10 Retrieving
70 mm 45 250 mL Laboratory GL Thread Bottle, With Cap, Safety Coated 5539-115 146 mm 89051-786 Case of 10 Retrieving
86 mm 45 500 mL Laboratory GL Thread Bottle, With Cap, Safety Coated 5539-118 183 mm 89051-788 Case of 10 Retrieving
101 mm 45 1 L Laboratory GL Thread Bottle, With Cap, Safety Coated 5539-122 230 mm 89051-790 Case of 10 Retrieving
136 mm 45 2 L Laboratory GL Thread Bottle, With Cap, Safety Coated 5539-125 267 mm 89051-792 Case of 10 Retrieving
186 mm 45 5 L Laboratory GL Thread Bottle, With Cap, Safety Coated 5539-130 340 mm 89051-795 Case of 1 Retrieving
56 mm 45 100 mL Laboratory GL Thread Bottle, Without Cap, Safety Coated 5539-150 108 mm 89199-030 Case of 10 Retrieving
70 mm 45 250 mL Laboratory GL Thread Bottle, Without Cap, Safety Coated 5539-152 146 mm 89199-032 Case of 10 Retrieving
86 mm 45 500 mL Laboratory GL Thread Bottle, Without Cap, Safety Coated 5539-154 183 mm 89199-034 Case of 10 Retrieving
101 mm 45 1 L Laboratory GL Thread Bottle, Without Cap, Safety Coated 5539-156 230 mm 89199-036 Case of 10 Retrieving
136 mm 45 2 L Laboratory GL Thread Bottle, Without Cap, Safety Coated 5539-158 267 mm 89199-038 Case of 10 Retrieving
186 mm 45 5 L Laboratory GL Thread Bottle, Without Cap, Safety Coated 5539-160 340 mm 89199-040 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Add to Shopping List
Existing List New List
Select List Name: Enter List Name: 
CALL VWR
1.800.932.5000