Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Revco® UxF Series Ultra-Low Temperature Freezers, Thermo Scientific

Supplier: Thermo Scientific

Revco® UxF ultra-low temperature freezers provide outstanding sample protection, while maintaining the highest standards in performance, energy efficiency and capacity per footprint.

  • Maximize sample storage capacity while minimizing the freezer's physical space
  • Easy-to-remove, washable filter provides protection against dust on the condense
  • Refrigeration system with high-efficiency compressors for excellent performance and reliability
Provides faster recovery after door openings, tighter uniformity, and reduced energy usage, achieved primarily through the use of a brazed plate heat exchanger for greater surface area and heat transfer.
The Revco® UxF Series has an innovative, touch-screen control panel can monitor the freezer’s health 24/7, set preferences and access a detailed event log. Event logs can be downloaded via the built-in USB port for freezer performance archiving. All Revco® UxF freezers feature a choice of energy-savings or high-performance operating modes allowing for energy savings and ultra-tight temperature uniformity.

Construction features include: five polystyrene insulated inner doors with embedded rare earth magnets; reinforced shelves that support up to 278 lbs. of cargo (depending on model); 4x7 outer door gasket with four points of security and seven zones of protection to prevent air migration, dual 2" lockable casters, brazed plate heat exchanger, surge suppression, voltage buck/boost, 2 1" access ports, door mounted pressure equalization port, Warranty: 2 year parts and labor, additional 3 year compressor parts only.

Backup systems inject liquid CO2 or LN2 when cabinet temperature warms to a preset level. Built-in backup systems are available on UxF400,500,600 and 700 models only; UxF300 series models require a freestanding backup system. Liquid flow shuts off automatically when the freezer door is opened. Rack solutions are available for all UxF models. Choose from both side access or sliding drawer racks for 2" boxes, 3" boxes, standard microplates, and deep well plates. Capacities vary by rack and freezer, please see ordering table below.

Optional proximity card locking option provides access control allowing for enhanced security. On-board computer records who opened the door, when it occurred and for how long. Temperature recorders are 15.2cm (6") circular models with 7-day temperature recording capability and use pressure-sensitive chart paper (sold separately) and do not require inking. The remote alarm module provides an audible and visual signal in the event of a temperature rise or power failure.

UL, cUL Listed - CE Market.

Ordering Information: Racks, back-up systems and other accessories sold separately. Revco® UxF freezers are supplied with manufacturer's two-year parts and labor warranty (five years in Canada), plus an additional three-year warranty on compressor parts. Upright freezers have five inner doors and include a four adjustable stainless steel shelves. All models include two 2.5cm (1") diameter access ports with caps. See 89200-252 for accessory listings. Contact your sales representative for further information on all optional accessories available.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Electrical Capacity Volume Exterior Dimensions Maximum Load per Shelf Shipping Weight Supplier No. VWR Catalog Number Unit Price Quantity
115V, 60Hz 300 Cryoboxes 422 L (14.9 cu. ft.) 198.1H x 68.6W x 95.5D cm (78 x 27 x 373/5") 56.8 kg (125 lbs.) 303 kg (668 lbs.) UXF30086A 89194-946 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
115V, 60Hz 600 Cryoboxes 815 L (28.8 cu. ft.) 198.1H x 110.8W x 95.5D cm (78 x 433/5 x 373/5") 110.1 kg (245 lbs.) 388 kg (855 lbs.) UXF60086A 89194-964 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
115V, 60Hz 500 Cryoboxes 680 L (24 cu. ft.) 198.1H x 96.5W x 95.5D cm (78 x 38 x 373/5") 91.7 kg (205 lbs.) 355 kg (783 lbs.) UXF50086A 89194-958 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
115V, 60Hz 400 Cryoboxes 549 L (19.4 cu. ft.) 198.1H x 82.2W x 95.5D cm (78 x 322/5 x 373/5") 73.4 kg (165 lbs.) 332 kg (731 lbs.) UXF40086A 89194-952 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
115V, 60Hz 500 Cryoboxes 680 L (24 cu. ft.) - - - UXF50086ARAK 89200-430 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 300 Cryoboxes 419 L (14.8 cu. ft.) - - - UXF30086ARAK 89200-418 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 300 Cryoboxes 419 L (14.8 cu. ft.) - - - UXF30086A5BB 89209-010 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 400 Cryoboxes 547 L (19.3 cu. ft.) - - - UXF40086A5BB 89209-014 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 500 Cryoboxes 680 L (24 cu. ft.) 198.1H x 96.5W x 95.5D cm (78 x 38 x 37.6") - 355 kg (782 lbs.) UXF50086A5BB 89209-018 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 600 Cryoboxes 810 L (28.6 cu. ft.) - - - UXF60086A5BB 89209-022 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 600 Cryoboxes 810 L (28.6 cu. ft.) - - - UXF60086ARAK 89200-436 Each Retrieving
Product available on GSA Advantage®
115V, 60Hz 400 Cryoboxes 547 L (19.3 cu. ft.) - - - UXF40086ARAK 89200-424 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 600 Cryoboxes 815 L (28.8 cu. ft.) 198.1H x 110.8W x 95.5D cm (78 x 433/5 x 373/5") 110.1 kg (245 lbs.) 388 kg (855 lbs.) UXF60086D 89194-966 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
208/230V, 60Hz 600 Cryoboxes 810 L (28.6 cu. ft.) - - - UXF60086DRAK 89200-438 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 300 Cryoboxes 422 L (14.9 cu. ft.) 198.1H x 68.6W x 95.5D cm (78 x 27 x 373/5") 56.8 kg (125 lbs.) 303 kg (668 lbs.) UXF30086D 89194-948 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
208/230V, 60Hz 400 Cryoboxes 549 L (19.4 cu. ft.) 198.1H x 82.2W x 95.5D cm (78 x 322/5 x 373/5") 73.4 kg (165 lbs.) 332 kg (731 lbs.) UXF40086D 89194-954 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
208/230V, 60Hz 700 Cryoboxes 949 L (33.5 cu. ft.) 198.1H x 125.1W x 95.5D cm (78 x 491/5 x 373/5") 128.4 kg (285 lbs.) 432 kg (952 lbs.) UXF70086D 89194-970 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
208/230V, 60Hz 500 Cryoboxes 680 L (24 cu. ft.) 198.1H x 96.5W x 95.5D cm (78 x 38 x 373/5") 91.7 kg (205 lbs.) 355 kg (783 lbs.) UXF50086D 89194-960 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
208/230V, 60Hz 700 Cryoboxes 943 L (33.3 cu. ft.) - - - UXF70086DRAK 89200-442 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 500 Cryoboxes 680 L (24 cu. ft.) - - - UXF50086DRAK 89200-432 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 400 Cryoboxes 547 L (19.3 cu. ft.) - - - UXF40086DRAK 89200-426 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 300 Cryoboxes 419 L (14.8 cu. ft.) - - - UXF30086DRAK 89200-420 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 300 Cryoboxes 419 L (14.8 cu. ft.) - - - UXF30086D5BB 89209-012 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 400 Cryoboxes 547 L (19.3 cu. ft.) - - - UXF40086D5BB 89209-016 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 500 Cryoboxes 680 L (24 cu. ft.) - - - UXF50086D5BB 89209-020 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 600 Cryoboxes 810 L (28.6 cu. ft.) - - - UXF60086D5BB 89209-024 Each Retrieving
Product available on GSA Advantage®
208/230V, 60Hz 700 Cryoboxes 949 L (33.5 cu. ft.) 198.1H x 125.1W x 95.5D cm (78 x 491/5 x 373/5") - 432 kg (951 lbs.) UXF70086D5BB 89209-026 Each Retrieving
Product available on GSA Advantage®
230V, 50Hz 700 Cryoboxes 949 L (33.5 cu. ft.) 198.1H x 125.1W x 95.5D cm (78 x 491/5 x 373/5") 128.4 kg (285 lbs.) 432 kg (952 lbs.) UXF70086V 89194-972 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
230V, 50Hz 500 Cryoboxes 680 L (24 cu. ft.) 198.1H x 96.5W x 95.5D cm (78 x 38 x 373/5") 91.7 kg (205 lbs.) 355 kg (783 lbs.) UXF50086V 89194-962 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
230V, 50Hz 600 Cryoboxes 815 L (28.8 cu. ft.) 198.1H x 110.8W x 95.5D cm (78 x 433/5 x 373/5") 110.1 kg (245 lbs.) 388 kg (855 lbs.) UXF60086V 89194-968 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
230V, 50Hz 300 Cryoboxes 422 L (14.9 cu. ft.) 198.1H x 68.6W x 95.5D cm (78 x 27 x 373/5") 56.8 kg (125 lbs.) 303 kg (668 lbs.) UXF30086V 89194-950 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
230V, 50Hz 400 Cryoboxes 549 L (19.4 cu. ft.) 198.1H x 82.2W x 95.5D cm (78 x 322/5 x 373/5") 73.4 kg (165 lbs.) 332 kg (731 lbs.) UXF40086V 89194-956 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories for Revco® UxF Series Ultra-Low Temperature Freezers, Thermo Scientific
Description Supplier No. VWR Catalog Number Unit Price Quantity
- FCRP567R 89210-956 Each Retrieving
Product available on GSA Advantage®
- RSK28CFSD5 89239-470 Each Retrieving
Product available on GSA Advantage®
Access Card Option, Field Installed for UxF30086, UxF40086, UxF50086, UxF60086, UxF70086 FRAC34567 89200-268 Each Retrieving
Product available on GSA Advantage®
Accessory Key Pack for Access Control Option compatible with Revco UxF ACE34567 89200-248 Each Retrieving
Product available on GSA Advantage®
Chart Recorder, Factory Installed for Revco UxF40086, UxF50086, UxF60086, UxF70086 CR4567 89200-256 Each Retrieving
Product available on GSA Advantage®
Chart Recorder, Field Installed for Revco UxF400 (all voltages) FCR400R 89200-232 Each Retrieving
Product available on GSA Advantage®
Chart Recorder, Field Installed for Revco UxF500, UxF600, UxF700 (all voltages) FCR567R 89200-234 Each Retrieving
Product available on GSA Advantage®
CO2 Backup System, 115V/60Hz, Field Installed for Revco UxF30086A FCO300A 89200-222 Each Retrieving
Product available on GSA Advantage®
CO2 Backup System, 230V/50Hz, Euro Plug, Field Installed for Revco UxF30086V FCO300V 89200-226 Each Retrieving
Product available on GSA Advantage®
CO2 Backup System, 230V/50 Hz, UK Plug,Field Installed for Revco UxF30086W FCO300W 89200-228 Each Retrieving
Product available on GSA Advantage®
CO2 Backup System, 230V/60Hz, Field Installed for Revco UxF30086D FCO300D 89200-224 Each Retrieving
Product available on GSA Advantage®
LN2 Backup System, 115V/60Hz, Field Installed for Revco UxF30086A FLN300A 89200-212 Each Retrieving
Product available on GSA Advantage®
LN2 Backup System, 230V/50Hz, Euro Plug, Field Installed for Revco UxF30086V FLN300V 89200-216 Each Retrieving
Product available on GSA Advantage®
LN2 Backup System, 230V/50 Hz, UK Plug,Field Installed for Revco UxF30086W FLN300W 89200-218 Each Retrieving
Product available on GSA Advantage®
LN2 Backup System, 230V/60Hz, Field Installed for Revco UxF30086D FLN300D 89200-214 Each Retrieving
Product available on GSA Advantage®
Shelf Kit for Revco UxF30086 SK300 89200-236 Each Retrieving
Product available on GSA Advantage®
Shelf Kit for Revco UxF40086 SK400 89200-238 Each Retrieving
Product available on GSA Advantage®
Shelf Kit for Revco UxF50086 SK500 89200-240 Each Retrieving
Product available on GSA Advantage®
Shelf Kit for Revco UxF60086 SK600 89200-242 Each Retrieving
Product available on GSA Advantage®
Shelf Kit for Revco UxF70086 SK700 89200-244 Each Retrieving
Product available on GSA Advantage®
Side Access Microplate Rack standard/deepwell, 5 inner door freezers, fits 28cf, 32cf, 300, 400, 500, 600, 700 5.5 x 9.4 x 25.7 398322 89200-164 Each Retrieving
Side Access rack 2" box, 5 inner door freezers 20 bxs/rack, fits 300, 400, 500, 600, 700 uprights 5.4 x 9.4 x 27.5 1950518 89200-198 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty