Would you like to log in again?
96-Well Microtitration Plates, Corning®
Supplier: Corning
Ideal for most commonly used cell based and biochemical assays.
Choose from flat-, round-, or V-bottom configuration in medical grade polystyrene or flexible vinyl (polyvinyl chloride). Plates are available tissue culture treated, untreated, or with nonbinding surface (NBS®) for reduced non-specific protein and nucleic acid binding. Plates are available in sterile and nonsterile formats.
The sterile universal lid, an accessory for Corning® 96-Well Surface-Modified Microplates, is made to fit all assay plates and blocks. Lid features a robotic-friendly design and a shorter overhang for ease with bar code readers. The Acetate Sealing Tape, Precut, Nonsterile, All Plates, also an accessory for the Corning® 96-Well Surface-Modified Microplates is precut and pressure sensitive.
Ordering Information: The lids for 96-well plates fit all plates except the Round-Bottom, Flexible Vinyl plate and the V-Bottom, Flexible Vinyl plate. Both of the Sterile Flat-Bottom Plate with Low Evaporation Lid options have special low evaporation lids. Lids are sold separately.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Material Well Volume Surface Treatment Well Shape Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Nonsterile Plate without Lid Flexible Vinyl 250 µL Untreated Round-Bottom 25/Bag 2797 29442-384 Case of 100 Retrieving
Nonsterile Plate without Lid Flexible Vinyl 260 µL Untreated V-Bottom 25/Bag 2897 29442-386 Case of 100 Retrieving
Nonsterile Plate without Lid Polystyrene, Improved Hydrophilicity 360 µL Untreated Round-Bottom 25/Bag 3798 29442-398 Case of 100 Retrieving
Nonsterile Plate without Lid Polystyrene, Improved Hydrophilicity 320 µL Untreated V-Bottom 25/Bag 3898 29442-404 Case of 100 Retrieving
Nonsterile Plate without Lid Polystyrene 360 µL Untreated Round-Bottom 25/Bag 3797 29442-396 Case of 100 Retrieving
Nonsterile Plate without Lid Flexible Vinyl 250 µL Untreated Flat-Bottom 25/Bag 2595 29442-306 Case of 100 Retrieving
Nonsterile Plate without Lid Polystyrene 360 µL Untreated Flat-Bottom 25/Bag 9017 29442-070 Case of 100 Retrieving
Nonsterile Plate without Lid Polystyrene 320 µL Untreated V-Bottom 25/Bag 3897 29442-402 Case of 100 Retrieving
Nonsterile Plate without Lid Polystyrene 360 µL Nonbinding Flat-Bottom 25/Bag 3641 29445-142 Case of 100 Retrieving
Sterile Flat-Bottom Plate with Lid Polystyrene 360 µL Tissue Culture Treated Flat-Bottom 5/Bag 3598 29442-058 Case of 100 Retrieving
Sterile Flat-Bottom Plate with Lid Polystyrene 360 µL Tissue Culture Treated Flat-Bottom Individually Wrapped 3599 29442-060 Case of 100 Retrieving
Sterile Flat-Bottom Plate with Lid Polystyrene 360 µL Tissue Culture Treated Flat-Bottom 10/Bag 3997 29443-954 Case of 50 Retrieving
Sterile Flat-Bottom Plate with Low Evaporation Lid Polystyrene 360 µL Tissue Culture Treated Flat-Bottom 5/Bag 3585 29442-050 Case of 50 Retrieving
Sterile Flat-Bottom Plate with Low Evaporation Lid Polystyrene 360 µL Tissue Culture Treated Flat-Bottom Individually Wrapped 3595 29442-054 Case of 50 Retrieving
Sterile Plate with Lid Polystyrene 360 µL Untreated Flat-Bottom 20/Bag 3370 25381-056 Case of 100 Retrieving
Sterile Plate without Lid Polystyrene 330 µL Tissue Culture Treated Round-Bottom 25/Bag 3360 29444-454 Case of 100 Retrieving
Sterile Plate without Lid Polystyrene 320 µL Untreated V-Bottom Individually Wrapped 3896 29442-400 Case of 48 Retrieving
Sterile Plate without Lid Polystyrene 360 µL Untreated Round-Bottom Individually Wrapped 3367 29444-288 Case of 50 Retrieving
Sterile Plate without Lid Polystyrene 360 µL Untreated Round-Bottom 25/Bag 3795 29442-392 Case of 100 Retrieving
Sterile Plate without Lid Polystyrene 330 µL Untreated Round-Bottom 20/Bag 3788 29445-154 Case of 100 Retrieving
Sterile Round-Bottom Plate with Lid Polystyrene 330 µL Tissue Culture Treated Round-Bottom Individually Wrapped 3799 29442-066 Case of 50 Retrieving
Sterile V-Bottom Plate with Lid Polystyrene 380 µL Tissue Culture Treated V-Bottom Individually Wrapped 3894 29442-068 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per  Qty