Your session will expire in 15 minutes.
Would you like to keep your session alive?
Would you like to log in again?
VWR® Test Tubes, Borosilicate Glass
Supplier: VWR International
Made from Type I, Class A borosilicate glass, tubes feature a sturdy lip and uniform wall thickness for maximum heat transfer and chemical resistance. Durable white ceramic enamel marking spot.
Meet ASTM Specification E982, Type I requirements.
Log in to see your contract pricing and availability.
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Volume Tube Dimensions VWR Catalog Number Unit Price Quantity
16 mL 15 x 125 mm 89000-482 Case of 720 Retrieving Purchase Hold
Pack of 72 Retrieving Purchase Hold
24 mL 16 x 150 mm 89000-484 Case of 576 Retrieving Purchase Hold
Pack of 72 Retrieving Purchase Hold
28 mL 18 x 150 mm 89000-486 Case of 576 Retrieving Unavailable
Pack of 72 Retrieving Unavailable
36 mL 20 x 150 mm 89000-488 Case of 576 Retrieving Unavailable
Pack of 72 Retrieving Purchase Hold
4 mL 10 x 75 mm 89000-476 Case of 720 Retrieving Purchase Hold
Pack of 72 Retrieving Purchase Hold
6 mL 12 x 75 mm 89000-478 Case of 720 Retrieving Unavailable
Pack of 72 Retrieving Purchase Hold
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
purchaseHold Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per  Qty