Would you like to log in again?
ACTERO™ E. coli Enrichment Media, FoodChek Systems
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing enabling target pathogen present to grow and be detected in test samples. The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ STEC Enrichement Media is used for the selective growth of Shiga-Toxin producing E. coli (STEC). It has been AOAC-PTM validated to be used in raw ground beef.
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Elite Enrichment Media
Description Application Includes Yield Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ Modified Tryptone Soya Broth (MTSB) with E. coli 0157 Supplement Vials (VCC) Enrichment of Escherichia coli O157 from Food 500 g Dehydrated Media and 2 Vials of VCC Media 15 L Rehydrated Media FCM-008 97068-832 Each (500g) Retrieving Restricted
Actero™ E. coli 0157 Supplement Vials (VCC) Additional Selectivity for Enrichment Media 2 Vials of VCC Media 10 Tests per Vial FCM-002 89498-636 Pack of 2 Retrieving Restricted
Actero™ Modified Tryptone Soya Broth (MTSB) Enrichment of Escherichia coli O157 from Food (requires FCM-002 VCC supplement for selective enrichment) 500 g Dehydrated Media 15 L Rehydrated Media FCM-001 97068-828 Each (500g) Retrieving Restricted
Actero™ Modified Tryptone Soya Broth (MTSB) Media Pouches Enrichment of Escherichia coli O157 from Food 20 x 24.75 g Pouches of Dehydrated Media 20 Tests with 375 g Samples FCM-003 97068-834 Pack of 20 (750ml) Retrieving Restricted
Actero™ Shiga Toxin-Producing E. coli (STEC) Enrichment Media Selective Growth of STEC; Promotes Faster Selective Growth and Enrichment of STEC Strains; Validated to Performance Tested Methods of AOAC-RI in Ground Beef 500 g Dehydrated Media and 3 Vials of Supplement Media 30 L Rehydrated Media FCM-013 97068-840 Each (500g) Retrieving Restricted
Actero™ Salmonella/Shiga Toxin-Producing E. coli (STEC) Enrichment Media Selective Enrichment of Salmonella spp. in Food and on Environmental Surfaces; Enrichment of STEC Along or Co-Enriched with Salmonella in Ground Beef 500 g Dehydrated Media and 3 Vials of Supplement Media 30 L Rehydrated Media FCM-010 97068-838 Each (500g) Retrieving Restricted
Universal Enrichment Media
Description Application Includes Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ MacConkey Agar Isolation and Differentiation of Coliforms and Enteric Pathogens based upon the Fermentation of Lactose 2 kg Dehydrated Media FCM-132 10146-684 Each (2kg) Retrieving Restricted
Actero™ MacConkey Agar Isolation and Differentiation of Coliforms and Enteric Pathogens based upon the Fermentation of Lactose 10 kg Dehydrated Media FCM-131 10146-682 Each (10kg) Retrieving Restricted
Actero™ MacConkey Agar Isolation and Differentiation of Coliforms and Enteric Pathogens based upon the Fermentation of Lactose 500 g Dehydrated Media FCM-133 10146-686 Each (500g) Retrieving Restricted
Actero™ MacConkey Broth Cultivation of Gram Negative Lactose Fermenting Enteric Bacteria in Samples of Water, Milk and Food 500 g Dehydrated Media FCM-175 10158-896 Each (500g) Retrieving Restricted
Actero™ MacConkey Broth Cultivation of Gram Negative Lactose Fermenting Enteric Bacteria in Samples of Water, Milk and Food 10 kg Dehydrated Media FCM-173 10158-892 Each (10kg) Retrieving Restricted
Actero™ MacConkey Broth Cultivation of Gram Negative Lactose Fermenting Enteric Bacteria in Samples of Water, Milk and Food 2 kg Dehydrated Media FCM-174 10158-894 Each (2kg) Retrieving Restricted
Actero™ MacConkey Sorbitol Agar Isolation and Detection of Pathogenic E. coli. 2 kg Dehydrated Media FCM-177 10158-900 Each (2kg) Retrieving Restricted
Actero™ MacConkey Sorbitol Agar Isolation and Detection of Pathogenic E. coli. 500 g Dehydrated Media FCM-178 10158-902 Each (500g) Retrieving Restricted
Actero™ MacConkey Sorbitol Agar Isolation and Detection of Pathogenic E. coli. 10 kg Dehydrated Media FCM-176 10158-898 Each (10kg) Retrieving Restricted
Actero™ Modified EC Medium With the Addition of Novobiocin for the Pre-Enrichment of E. coli 0157:H7 10 kg Dehydrated Media FCM-094 10066-042 Each (10kg) Retrieving Restricted
Actero™ Modified EC Medium With the Addition of Novobiocin for the Pre-Enrichment of E. coli 0157:H7 2 kg Dehydrated Media FCM-095 10066-044 Each (2kg) Retrieving Restricted
Actero™ Modified EC Medium With the Addition of Novobiocin for the Pre-Enrichment of E. coli 0157:H7 500 g Dehydrated Media FCM-096 10066-046 Each (500g) Retrieving Restricted
Actero™ Violet Red Bile Agar with MUG (VRBA w/MUG) Fluorogenic Eetection of Escherichia coli in Food, Milk, Water and Sanitary Materials; Detects Growth of Lactose Fermenting Coliforms. 2 kg Dehydrated Media FCM-077 10066-024 Each (2kg) Retrieving Restricted
Actero™ Violet Red Bile Agar with MUG (VRBA w/MUG) Fluorogenic Eetection of Escherichia coli in Food, Milk, Water and Sanitary Materials; Detects Growth of Lactose Fermenting Coliforms. 10 kg Dehydrated Media FCM-076 10066-022 Each (10kg) Retrieving Restricted
Actero™ Violet Red Bile Agar with MUG (VRBA w/MUG) Fluorogenic Eetection of Escherichia coli in Food, Milk, Water and Sanitary Materials; Detects Growth of Lactose Fermenting Coliforms. 500 g Dehydrated Media FCM-078 10066-026 Each (500g) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty