Would you like to log in again?
ACTERO™ Salmonella Enrichment Media, FoodChek Systems
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing enabling target pathogen present to grow and be detected in test samples. The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ Salmonella Enrichement Media recieved the AOAC-PTM (041403) accreditation to be used as single-step recover and enrichment of Salmonella spp. from selected food and environmental samples.
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Elite Enrichment Media
Description Application Yield Includes Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ Salmonella Enrichment Media Improved Enrichment of Salmonella spp. from Food and Environmental Surfaces; Validated to Performance Tested Methods of AOAC-RI in Food and Environmental Surfaces 30 L Rehydrated Media 500 g Dehydrated Media and 3 Vials of Supplement Media FCM-009 97068-836 Each (500g) Retrieving Restricted
Actero™ Buffered Peptone Water Pre-enriching Damaged Salmonella spp. from Various Food Sources - 200 g MediaPouch FCM-191 10142-750 Case of 50 (200 g) Retrieving Restricted
Each(200g) Retrieving Restricted
Actero™ Buffered Peptone Water Pre-enriching Damaged Salmonella spp. from Various Food Sources 20 L Rehydrated Media 25 x 200 g Dehydrated Media FCM-194 10153-806 Case of 25 (400 g) Retrieving Restricted
Each(400g) Retrieving Restricted
Universal Enrichment Media
Description Application Includes Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ Bismuth Sulfite Agar Isolation of Salmonella spp. Particularly Isolation of Salmonella typhi from Clinical and Environmental Specimens 10 kg Dehydrated Media FCM-088 10066-030 Each (10kg) Retrieving Restricted
Actero™ Bismuth Sulfite Agar Isolation of Salmonella spp. Particularly Isolation of Salmonella typhi from Clinical and Environmental Specimens 500 g Dehydrated Media FCM-090 10066-034 Each (500g) Retrieving Restricted
Actero™ Bismuth Sulfite Agar Isolation of Salmonella spp. Particularly Isolation of Salmonella typhi from Clinical and Environmental Specimens 2 kg Dehydrated Media FCM-089 10066-032 Each (2kg) Retrieving Restricted
Actero™ Brilliant Green Agar Isolation of Salmonella spp. 10 kg Dehydrated Media FCM-140 10146-700 Each (10kg) Retrieving Restricted
Actero™ Brilliant Green Agar Isolation of Salmonella spp. 2 kg Dehydrated Media FCM-141 10146-702 Each (2kg) Retrieving Restricted
Actero™ Brilliant Green Agar Isolation of Salmonella spp. 500 g Dehydrated Media FCM-142 10146-704 Each (500g) Retrieving Restricted
Actero™ Brilliant Green Bile Broth, 2% Confirming the Presence of Coliforms in Water, Sewage and Dairy 10 kg Dehydrated Media FCM-188 10158-922 Each (10kg) Retrieving Restricted
Actero™ Brilliant Green Bile Broth, 2% Confirming the Presence of Coliforms in Water, Sewage and Dairy 2 kg Dehydrated Media FCM-189 10158-924 Each (2kg) Retrieving Restricted
Actero™ Brilliant Green Bile Broth, 2% Confirming the Presence of Coliforms in Water, Sewage and Dairy 500 g Dehydrated Media FCM-190 10158-926 Each (500g) Retrieving Restricted
Actero™ Buffered Peptone Water Pre-Enriching Damaged Salmonella spp.from Food 2 kg Dehydrated Media FCM-065 10066-058 Each (2kg) Retrieving Restricted
Actero™ Buffered Peptone Water Pre-Enriching Damaged Salmonella spp.from Food 10 kg Dehydrated Media FCM-064 10066-056 Each (10kg) Retrieving Restricted
Actero™ Buffered Peptone Water Pre-Enriching Damaged Salmonella spp.from Food 500 g Dehydrated Media FCM-066 10066-060 Each (500g) Retrieving Restricted
Actero™ Buffered Peptone Water, ISO Pre-Enriching Damaged Salmonella spp.from Food 500 g Dehydrated Media FCM-148 10146-716 Each (500g) Retrieving Restricted
Actero™ Buffered Peptone Water, ISO Pre-Enriching Damaged Salmonella spp.from Food 10 kg Dehydrated Media FCM-146 10146-712 Each (10kg) Retrieving Restricted
Actero™ Buffered Peptone Water, ISO Pre-Enriching Damaged Salmonella spp.from Food 2 kg Dehydrated Media FCM-147 10146-714 Each (2kg) Retrieving Restricted
Actero™ Lactose Broth Detection and/or Verification of the Presence of Salmonella and other Coliforms in Water, Foods, Dairy, and Pharmaceuticals 10 kg Dehydrated Media FCM-049 10066-054 Each (10kg) Retrieving Restricted
Actero™ Lactose Broth Detection and/or Verification of the Presence of Salmonella and other Coliforms in Water, Foods, Dairy, and Pharmaceuticals 2 kg Dehydrated Media FCM-050 10066-108 Each (2kg) Retrieving Restricted
Actero™ Lactose Broth Detection and/or Verification of the Presence of Salmonella and other Coliforms in Water, Foods, Dairy, and Pharmaceuticals 500 g Dehydrated Media FCM-051 10066-110 Each (500g) Retrieving Restricted
Actero™ M Broth Cultivating Salmonella spp. in Foods and Feeds by the Accelerated EnrichmentSserology (ES) Procedure; Conforms to Recommended Procedures in Food Testing and Enzyme Immunoassays 2 kg Dehydrated Media FCM-086 10066-076 Each (2kg) Retrieving Restricted
Actero™ M Broth Cultivating Salmonella spp. in Foods and Feeds by the Accelerated EnrichmentSserology (ES) Procedure; Conforms to Recommended Procedures in Food Testing and Enzyme Immunoassays 500 g Dehydrated Media FCM-087 10066-078 Each (500g) Retrieving Restricted
Actero™ M Broth Cultivating Salmonella spp. in Foods and Feeds by the Accelerated EnrichmentSserology (ES) Procedure; Conforms to Recommended Procedures in Food Testing and Enzyme Immunoassays 10 kg Dehydrated Media FCM-085 10066-074 Each (10kg) Retrieving Restricted
Actero™ Tetrathionate Broth Used as a selective enrichment for the cultivation of Salmonella species, compete with intestinal flora 10 kg Dehydrated Media FCM-107 10500-800 Each (10kg) Retrieving Restricted
Actero™ Tetrathionate Broth Used as a selective enrichment for the cultivation of Salmonella species, compete with intestinal flora 500 kg Dehydrated Media FCM-109 10500-804 Each (500g) Retrieving Restricted
Actero™ Universal Pre-Enrichment Broth Recovery of Salmonella and Listeria from Food 10 kg Dehydrated Media FCM-143 10146-706 Each (10kg) Retrieving Restricted
Actero™ Universal Pre-Enrichment Broth Recovery of Salmonella and Listeria from Food 500 g Dehydrated Media FCM-145 10146-710 Each (500g) Retrieving Restricted
Actero™ Universal Pre-Enrichment Broth Recovery of Salmonella and Listeria from Food 2 kg Dehydrated Media FCM-144 10146-708 Each (2kg) Retrieving Restricted
Clamp Red Tubing Used for Manually Regulating Media Flow from the MediaBox Tubing Pack of 12 FCLM-033 10500-812 Pack of 12 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty