Would you like to log in again?
FoodChek™ Systems Listeria spp. Environmental Rapid Tests, FoodChek Systems Inc.
Supplier: Foodchek
The revolutionary new FoodChek™ Listeria spp. test is the fastest and most accurate test for detecting Listeria spp. from environmental samples (swabs) available in the marketplace today. It is easy-to-use, cost effective and increases both the profitiability and confidence in a processor's operational system.
FoodChek™ Listeria spp. is more sensitive than PCR systems, delivering accurate results in less than 24 hours including the sample enrichment development phase. Other competitive lab systems require 24 to 48 hours for the enrichment phase alone.
FoodChek™ Listeria spp. is significantly simpler to run than a typical microbial test in a laboratory setting as there is no need for expensive equipment or highly trained microbiologists. An in-house Good Laboratory Practice (GLP) technician is all that is required to perform the FoodChek™ test.
Using Actero™ Enrichment Media significantly reduces all sample enrichment phase timelines regardless of the type of “food pathogen testing system,” such as lateral flow, ELISA and PCR platforms. Enrichment time can be drastically improved by up to 30% faster for E. coli and Listeria, and up to 70% faster for Salmonella.
Ordering Information: Assay Cassettes can only be read on the FoodChek ™ MICT v2 Reader (sold separately). Actero™ Enrichment Media is sold separately from the FoodChek™ MICT Reader and Assay Cassettes.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Includes Supplier No. VWR Catalog Number Unit Price Quantity
FoodChek™ Listeria spp. Assay Cassettes 20 Listeria spp. Assay Cassettes FCLS-003 97068-826 Pack of 20 Retrieving Restricted
FoodChek™ Listeria spp. Sampling Swabs 100 FoodChek™ Long-Handled Sampling Swabs and 100 Vials of D/E Neutralizing Buffer FCLS-005 89498-634 Each Retrieving
Listeria spp. Sampling Kit 1 Kit of 20 Assay Cassettes, 20 Sani-Sticks, 20 D/E Neutralizer, 1 Cleaning Cassette FCLS-006 10031-444 Each Retrieving Restricted
Listeria spp. Sampling Kit (Bulk) 5 Kits of 20 Assay Cassettes, 20 Sani-Sticks, 20 D/E Neutralizer, 1 Cleaning Cassette (100 pcs) FCLS-007 10031-446 Pack of 5 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty