Would you like to log in again?
FoodChek™ Systems E. coli O157 Rapid Tests
Supplier: FOODCHEK SYSTEMS INC.
The revolutionary new FoodChek™ - E. coli 0157 test is the fastest and most accurate same-production shift test for E. coli O157 available in the marketplace today. It is easy-to-use, cost effective and increases both the profitability and confidence in a processor’s operational system
FoodChek™ - E. coli O157 is 50 times more sensitive than PCR systems, making it the world’s fastest E. coli O157 pathogen test and delivers accurate results in less than 8 hours including the sample enrichment development phase versus other current competitive lab tests that take from 12 to 24 hours for the enrichment phase alone.
FoodChek™ - E. coli O157 is significantly simpler to run than a typical microbial test in a laboratory setting as there is no need for expensive equipment and highly trained microbiologists. An in-house Good Laboratory Practice (GLP) technician is all that is required to perform the FoodChek™ test.
Assay Cassettes can only be read on the FoodChek ™ - MICT v2 Reader (Sold Separately).
Using Actero™ Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system" - such as lateral flow, ELISA and PCR platforms. Enrichment time can be drastically improved by up to 30% faster for E. coli and Listeria, and up to 70% faster for Salmonella. Actero™ Enrichment Media is sold separately from the FoodChek™ MICT Reader and Assay Cassettes.
AOAC Approved.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Includes Supplier No. VWR Catalog Number Unit Price Quantity
FoodChek™ E. coli O157 Assay Cassettes 1 Box of FoodChek™ E. coli O157 Assay Cassettes contains 20 Individual E. coli O157 Assay Cassettes FCEC-006 97068-824 Pack of 20 Retrieving Restricted
FoodChek™ E. coli O157 Bulk Media Test Kit The FoodChek™ E. coli O157 Bulk Media Test Kit contains: 20 Assay Cassettes, 2 vcc Supplement Vials, 500g Actero mTSB Media (dehydrated media, enough for 20 tests @ 375g samples) FCEC-002 97068-814 Case of 1 Retrieving Restricted
FoodChek™ E. coli O157 Carcass Sponge Test Kit The FoodChek™ E. coli O157 Carcass Sponge Test Kit contains: 500g Actero™ mTSB Media (dehydrated), 2 vcc upplement Vials, 20 Assay Cassettes and 20 Beef Carcass Swabs and Sample Bags FCEC-005 97068-818 Case of 1 Retrieving Restricted
FoodChek™ E. coli O157 Prepack Media Test Kit The FoodChek™ E. coli O157 Prepack Media Test Kit contains: 20 Assay Cassettes, 20 pre-packaged Actero™ mTSB pouches (dehydrated) and 2 vcc Supplement Vials (enough for 20 tests @ 375g samples) FCEC-004 97068-816 Case of 1 Retrieving Restricted
FoodChek™ E. coli O157 Test Kit The FoodChek™ E. coli O157 Test Kit contains: 20 Assay Cassettes and 2 vcc Supplement Vials (enough for 20 tests @ 375g samples) FCEC-001 97068-812 Case of 1 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty