Would you like to log in again?
FoodChek™ MICT v2 Reader, FoodChek Systems
Supplier: Foodchek
FoodChek’s MICT magnetic nanotechnology uses a compact desktop based diagnostic reading device (the “MICT Reader”) and reports the test results from disposable assay cassettes loaded with food samples that have been previously enriched for bacterial growth using another of FoodChek™’s products, the Actero™ Enrichment Media. The Patent Pending Actero™ Enrichment Media is used to incubate the food samples and enrich the growth of bacteria to allow the testing of pathogens common in foodborne diseases such as E. coli, Salmonella and Listeria.
The MICT Reader does not require a sophisticated technician in a commercial lab and is an automated, quantitative test that allows for tracking recording and printing of test results.
This allows for the fastest most accurate and easiest-to-use in-house pathogen testing systems in today's marketplace, is at least one order of magnitude more sensitive than PCR testing and a two order of magnitude higher than traditional (ELISA) tests and virtually eliminates human error.
FoodChek™'s MICT Cleaning Cassettes (sold separately) are specifically designed for use with it's MICT Readers. The cleaning cassettes are single use only.
FoodChek Systems Inc. recommends that the MICT reader should be cleaned after 20 tests or 1 month (whichever comes first) , in the event of an “invalid result”, when you are changing pathogen tests. (e.g.: E. coli to Listeria spp.).
FoodChek™'s MICT Verification Cassette must be run at the start of the testing day and is specifically designed for use with the FoodChek™ MICT Reader.
You can verify the FoodChek MICT Reader's accuracy and performance with the supplied verification cassette. This cassette contains three stable, calibrated test regions on one test strip. They are used to verify the instrument’s internal calibration (Accuracy) and performance (Noise).
The Verification Cassette is for multiple uses and only needs to be replaced when your VWR representative recommends to.
Ordering Information: All MICT Readers come with a power cord, power adaptor, 1 Stylus, CD User Manual, 3 rolls of thermo paper, 1 verification Cassette and a USB Memory Stick.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Electrical Supplier No. VWR Catalog Number Unit Price Quantity
FoodChek™ MICT Reader 110V FCR-004 97068-806 Each Retrieving
UL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories for FoodChek™ MICT v2 Reader, FoodChek Systems
Description Supplier No. VWR Catalog Number Unit Price Quantity
MICT Thermo Paper, 3 Rolls, Enough for approx. 100 Tests FCR-009 10031-442 Pack of 3 Retrieving Restricted
MICT Cleaning Cassettes FCR-002 97068-808 Pack of 10 Retrieving Purchase Hold
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty