Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Two-Sided Disposable Plastic Cuvettes

Supplier: VWR International

Cuvettes are constructed of high-quality polymethyl methacrylate (PMMA or "acrylic") or polystyrene (PS), and provide accurate, reliable results.

  • Compatible with most spectrophotometers and photometers
  • Ideal for most spectrophotometry applications in the visible and near-UV range
  • Cuvettes have a 10mm lightpath
  • Include an arrow mark to indicate direction of transmission and reduce variation
  • Available in macro and semi-macro sizes to accommodate most sample volumes
Acrylic cuvettes are suitable for assays between 300nm and 900nm. Polystyrene cuvettes are ideal for routine assays between 340nm and 900nm. They are grouped by manufacturing mold cavity number to ensure the lowest variation in extinction coefficient, and are packaged in low-dust, low-scratch material.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Material Volume Range VWR Catalog Number Unit Price Quantity
Semi-Micro Acrylic (PMMA) 1.5–3.0 mL 97000-590 Case of 500 Retrieving
Semi-Micro Polystyrene (PS) 1.5–3.0 mL 97000-586 Case of 500 Retrieving
Product available on GSA Advantage®
Macro Acrylic (PMMA) 2.5–4.5 mL 97000-588 Case of 500 Retrieving
Macro Polystyrene (PS) 2.5–4.5 mL 97000-584 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty