Would you like to log in again?
ACTERO™ Listeria Enrichment Media, FoodChek Systems
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing enabling targetl pathogen present to grow and be detected in test samples. The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ Listeria Enrichment Media recieved the AOAC-PTM (111201) accreditation to be used as a single-step recovery and enrichment of Listeria spp., including Listeria monocytogenes from selected food and environmental samples.
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Elite Enrichment Media
Description Application Includes Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ Listeria Enrichment Media Optimized for Single-Step Recovery and Enrichment of Listeria spp. from Food and Environmental Samples 500 g Dehydrated Media FCM-011 97068-830 Each (500g) Retrieving Restricted
Actero™ Listeria Enrichment Media Rapid Single-Step Enrichment of Listeria spp. from Food and Environmental Samples MediaPouch of 538 g to produce 10 L of Media FCM-192 10146-992 Case of 20 (538 g) Retrieving Restricted
Each(538g) Retrieving Restricted
Actero™ Listeria Enrichment Media Optimized for Single-Step Recovery and Enrichment of Listeria spp. from Food and Environmental Samples 2 kg Dehydrated Media FCM-022 10146-990 Case of 4 (2 kg) Retrieving Restricted
Each(2kg) Retrieving Restricted
Actero™ Listeria Enrichment Media Rapid Single-Step Enrichment of Listeria spp. from Food and Environmental Samples MediaPouch of 54 g to produce 1 L of Media FCM-193 10146-994 Case of 25 (54 g) Retrieving Restricted
Each(54g) Retrieving Restricted
Actero™ Listeria Enrichment Media Optimized for Single-Step Recovery and Enrichment of Listeria spp. from Food and Environmental Samples 10 Kg of Dehydrated Media FCM-023 103257-596 Each (10KU) Retrieving
Universal Enrichment Media
Description Application Includes Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ Listeria Enrichment Broth Listeria Enrichment Broth is used for the selective enrichment and isolation of Listeria spp. 10 kg Dehydrated Media FCM-037 89522-690 Each (5l) Retrieving Restricted
Actero™ Listeria Enrichment Broth Selective Enrichment and Isolation of Listeria spp. 2 kg Dehydrated Media FCM-038 89522-692 Each (10l) Retrieving Restricted
Actero™ Listeria Enrichment Broth Selective Enrichment and Isolation of Listeria spp. 500 g Dehydrated Media FCM-039 89522-694 Each (20l) Retrieving Restricted
Actero™ Buffered Listeria Enrichment Broth Selective enrichment of Listeria spp. 2 kg Dehydrated Media FCM-056 10065-990 Each (2kg) Retrieving Restricted
Actero™ Buffered Listeria Enrichment Broth Selective enrichment of Listeria spp. 500 g Dehydrated Media FCM-057 10065-992 Each (500g) Retrieving Restricted
Actero™ Buffered Listeria Enrichment Broth Selective enrichment of Listeria spp. 10 kg Dehydrated Media FCM-055 10065-988 Each (10kg) Retrieving Restricted
Actero™ Demi Fraser Broth Developed by Fraser and Sperber for the Rapid Detection of Listeria from Food and Environmental Samples 500 g of Dehydrated Media FCM-163 10146-670 Each (500g) Retrieving Restricted
Actero™ Demi Fraser Broth Developed by Fraser and Sperber for the Rapid Detection of Listeria from Food and Environmental Samples 2 kg of Dehydrated Media FCM-162 10146-668 Each (2kg) Retrieving Restricted
Actero™ Demi-Fraser Media (Modification of Fraser Broth) Rapid Detection of Listeria with the Addition of Ferric Ammonium Citrate 10 kg Dehydrated Media FCM-161 10146-654 Each (10kg) Retrieving Restricted
Actero™ Listeria Enrichment Broth Listeria Enrichment Broth is used for the selective enrichment and isolation of Listeria spp. 2 kg of Dehydrated Media FCM-138 10146-696 Each (2kg) Retrieving Restricted
Actero™ Listeria Enrichment Broth Listeria Enrichment Broth is used for the selective enrichment and isolation of Listeria spp. 500 g of Dehydrated Media FCM-139 10146-698 Each (500g) Retrieving Restricted
Actero™ Listeria Enrichment Broth Listeria Enrichment Broth is used for the selective enrichment and isolation of Listeria spp. 10 kg of Dehydrated Media FCM-137 10146-694 Each (10kg) Retrieving Restricted
Actero™ Oxford Listeria Agar With Addition of Antibiotics for the Selective Isolation of Listeria monocytogenes from Food 10 kg Dehydrated Media FCM-082 10066-068 Each (10kg) Retrieving Restricted
Actero™ Oxford Listeria Agar With Addition of Antibiotics for the Selective Isolation of Listeria monocytogenes from Food 2 kg Dehydrated Media FCM-083 10066-070 Each (2kg) Retrieving Restricted
Actero™ Oxford Listeria Agar With Addition of Antibiotics for the Selective Isolation of Listeria monocytogenes from Food 500 g Dehydrated Media FCM-084 10066-072 Each (500g) Retrieving Restricted
Actero™ Palcam Agar Detection of Listeria spp, Particularly Listeria monocytogene 10 kg Dehydrated Media FCM-100 10066-090 Each (10kg) Retrieving Restricted
Actero™ Palcam Agar Detection of Listeria spp, Particularly Listeria monocytogene 500 g Dehydrated Media FCM-102 10066-050 Each (500g) Retrieving Restricted
Actero™ Palcam Agar Detection of Listeria spp, Particularly Listeria monocytogene 2 kg Dehydrated Media FCM-101 10066-048 Each (2kg) Retrieving Restricted
Actero™ Palcam Broth With Supplements for the Selective Enrichment of Listeria spp. From Foods and Environmental Samples 2 kg Dehydrated Media FCM-104 10066-080 Each (2kg) Retrieving Restricted
Actero™ Palcam Broth With Supplements for the Selective Enrichment of Listeria spp. From Foods and Environmental Samples 10 kg Dehydrated Media FCM-103 10066-052 Each (10kg) Retrieving Restricted
Actero™ Palcam Broth With Supplements for the Selective Enrichment of Listeria spp. From Foods and Environmental Samples 500 g Dehydrated Media FCM-105 10066-082 Each (500g) Retrieving Restricted
Actero™ Palcam Supplement Detection and Isolation of Listeria Species When Incorporated into PALCAM Agar Palcam Broth 10 x 5 mL Vials FCM-211 10159-086 Pack of 10 (50ml) Retrieving Restricted
Actero™ Palcam Supplement Detection and Isolation of Listeria Species When Incorporated into PALCAM Agar Palcam Broth 60 mL Vial FCM-210 10159-084 Each (60ml) Retrieving Restricted
Actero™ Universal Pre-Enrichment Broth Recovery of Salmonella and Listeria from Food 10 kg Dehydrated Media FCM-143 10146-706 Each (10kg) Retrieving Restricted
Actero™ Universal Pre-Enrichment Broth Recovery of Salmonella and Listeria from Food 500 g Dehydrated Media FCM-145 10146-710 Each (500g) Retrieving Restricted
Actero™ Universal Pre-Enrichment Broth Recovery of Salmonella and Listeria from Food 2 kg Dehydrated Media FCM-144 10146-708 Each (2kg) Retrieving Restricted
Actero™ UVM Modified Listeria Enrichment Broth Selective Isolation and Enrichment of Listeria spp. from Clinical Specimens and Food 10 kg Dehydrated Media FCM-052 10065-982 Each (10kg) Retrieving Restricted
Actero™ UVM Modified Listeria Enrichment Broth Selective Isolation and Enrichment of Listeria spp. from Clinical Specimens and Food 500 g Dehydrated Media FCM-054 10065-986 Each (500g) Retrieving Restricted
Actero™ UVM Modified Listeria Enrichment Broth Selective Isolation and Enrichment of Listeria spp. from Clinical Specimens and Food 2 kg Dehydrated Media FCM-053 10065-984 Each (2kg) Retrieving Restricted
Supplement
Description Application Includes Supplier No. VWR Catalog Number Unit Price Quantity
Actero™ Oxford Listeria Supplement Detection and Isolation of Listeria Species When Incorporated into Listeria Oxford Agar 10 x 5 mL Vials FCM-213 10159-090 Pack of 10 (50ml) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty