Would you like to log in again?
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
These super absorbent bench protectors feature a two-layer, cushioned construction that is ideal for use in a laboratory or veterinary setting.
  • Two layers of protection
  • Leakproof backsheet
  • For laboratory and veterinary applications
  • Sold in rolls or as mats
  • Can be cut to desired size
The air-lay cellulose layer provides cushion for laboratory equipment and glassware to prevent breakage, while the leakproof plastic backsheet protects tables and benches from spills and moisture.
When these absorbent barriers are used in a veterinary setting, the absorbent material in these pads provides comfort for patients and can be used as a cage liner or table cover to absorb and contain moisture. Material can be cut to the desired size.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description W x L VWR Catalog Number Unit Price Quantity
Mat 45.7 x 50.8 cm (18 x 20") 89126-794 Case of 350 Retrieving
Roll 50.8 cm x 91.5 m (20" x 300') 89126-792 Case of 2 Retrieving
Roll 50.8 cm x 45.8 m (20" x 150') 89126-790 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty