Would you like to log in again?
VWR® Metal-Free Centrifuge Tubes, Polypropylene, Sterile
Supplier: VWR International
Ideal for environmental uses such as water analysis and other applications wherein tubes have the potential to contaminate samples.
  • Sterilized via irradiation
  • Conical shape
  • Polypropylene fabrication
Tubes are certified free of major critical trace metals to less than 1ppb. Conical centrifuge tubes are available in two sizes with natural flat caps for easy labeling. The polypropylene tubes and polyethylene caps are made of ultra-high purity CFR 21 compliant resins. Tubes feature printed graduation marks and a large white writing spot. All tubes have been sterilized via irradiation.
Ordering Information: Tubes packaged in bulk bags are supplied 50 tubes per bag, 10 bags per case. SBS recyclable rack packaged tubes are supplied 25 per rack, 2 racks per pack, 10 packs per case.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Volume Maximum RCF Packaging VWR Catalog Number Unit Price Quantity
15 mL 12,500 g SBS Recyclable Rack 89049-172 Case of 500 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Product is manufactured with low CO₂ impact Product available on GSA Advantage®
15 mL 12,500 g Bulk Bag 89049-170 Case of 500 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Product is manufactured with low CO₂ impact Product available on GSA Advantage®
50 mL 12,500 g Bulk Bag 89049-174 Case of 500 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Product is manufactured with low CO₂ impact Product available on GSA Advantage®
50 mL 12,500 g SBS Recyclable Rack 89049-176 Case of 500 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Product is manufactured with low CO₂ impact
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste, Product and/or packaging is safer to human and environmental health, Product is manufactured with low CO₂ impact
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty