Would you like to log in again?
VWR® Centrifuge Tubes with Flat Caps, Polypropylene, Sterile
Supplier: VWR International
Presterilized polypropylene tubes feature white graduations and flat high density polyethylene caps.
  • Natural coloring allows observation without being clear
  • Supplied with purple cap
Bulk packed bags simplify storage. Tubes provide good chemical and physical strength and lower protein binding than polystyrene tubes. Natural coloring allows fluid levels to be viewed, although tubes are not clear.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Volume Packaging VWR Catalog Number Unit Price Quantity
Centrifuge Tubes 15 mL 50/Bag 89004-368 Case of 500 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Product is manufactured with low CO₂ impact Product available on GSA Advantage®
Centrifuge Tubes 50 mL 50/Bag 89004-364 Case of 500 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Product is manufactured with low CO₂ impact Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste, Product and/or packaging is safer to human and environmental health, Product is manufactured with low CO₂ impact
Max. RCF 7500 g
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty