Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Falcon® Centrifuge Tubes, Polypropylene, Sterile, Corning®

Supplier: CORNING

These conical tubes are ideal for centrifugation and serial dilutions, as well as storage of critical laboratory reagents including culture media, buffers, and antibodies. They have bold blue printed graduations and a large white writing patch. The medical-style packaging assures sterile presentation. Polystyrene tube racks for 15mL tubes can be easily divided into two racks for versatility. Irradiation-sterilized. Nonpyrogenic. Non-cytotoxic.

Note: Tubes should not be frozen in racks.

Ordering Information: All tubes supplied with plug-seal screw caps. Package 62406-200 also includes one rack.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Volume Maximum RCF O.D. x L Graduations Packaging Supplier No. VWR Catalog Number Unit Price Quantity
15 mL 12,000 g 17 x 120 mm 0.5 mL 50/Bag 352196 62406-200 Case of 500 Retrieving
15 mL 12,000 g 17 x 120 mm 0.5 mL 50/Bag 352096 21008-936 Case of 500 Retrieving
15 mL 12,000 g 17 x 120 mm 0.5 mL 50/Rack 352097 21008-918 Case of 500 Retrieving
50 mL 16,000 g 30 x 115 mm 5 mL 25/Bag 352070 21008-940 Case of 500 Retrieving
50 mL 16,000 g 30 x 115 mm 5 mL 25/Rack 352098 21008-951 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Replacement Screw Caps for 50 mL Tubes 50/Bag 358206 12777-064 Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty