Would you like to log in again?
VWR® Aluminum Foils for PCR and Cold Storage
Supplier: VWR International
VWR® Aluminum Foils for PCR and Cold Storage provide a secure and reliable sealing area for all PCR plates.
  • 38µm thick aluminum foils
  • Equipped with strong, medical-grade adhesives
  • RNase-, DNase-, and nuclease-free
Soft, non-permeable 38µm thick aluminum foils with strong, medical-grade adhesives eliminate the need for heat-sealing devices or mats during thermal cycling. Each foil provides sufficient sealing area for all PCR plates. Aluminum foil film is ideal for PCR applications where piercing with pipet tips or robotic probes is required for product recovery. Unlike comparable foils, these foils exhibit less tendency to roll back on themselves when removing the backing paper and they conform to the plate during application. All foils are RNase-, DNase-, and nuclease-free. Dimensions: 14.2x8.2 cm (55/8x31/4"). Length with end tabs removed is 12.5 cm (415/16").
Sterile foils are packaged in tamper-evident bags of 25.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Sterility VWR Catalog Number Unit Price Quantity
Nonsterile 60941-074 Case of 100 Retrieving
Sterile 60941-076 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty