VWR® Pipet Filler, Synthetic Rubber
Supplier: VWR, part of Avantor
VWR®
K200-VWR
53502-205EA 54.95 USD
53502-205
VWR® Pipet Filler, Synthetic Rubber
Pipets Pipet Filler Bulbs
Pipet fillers do away with the frequently hazardous practice of drawing liquids into pipets by mouth.
  • No metal parts to corrode
For use with pipets from 2 to 50 ml. Made of synthetic rubber with glass ball valves.
Ordering information: Supplied with an adapter for capacities greater than 20 ml.