Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Laboratory Bottles, High-Density Polyethylene

Supplier: VWR International

Translucent, rigid bottles are for general laboratory use and for storing, shipping, and packing liquids.

  • Excellent chemical resistance
  • Durable for long-term use
  • High-density polyethylene construction
  • Translucent siding
Can be stored in freezer to -100°C (-148°F). Excellent chemical resistance to most acids, bases, and alcohols. Can be used with most corrosives. Durable enough for long-term use. Polypropylene screw closure included.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Capacity Cap Size VWR Catalog Number Unit Price Quantity
Narrow Mouth Bottles 4 mL (0.14 oz.) 13-415 414004-151 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 44928 Retrieving
Narrow Mouth Bottles 4 mL (0.14 oz.) 13-415 89221-580 Case of 2,000 Retrieving
Narrow Mouth Bottles 8 mL (0.27 oz.) 20-410 414004-152 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 21600 Retrieving
Narrow Mouth Bottles 8 mL (0.27 oz.) 20-410 89221-582 Case of 2,000 Retrieving
Narrow Mouth Bottles 15 mL (0.5 oz.) 20-410 414004-153 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 10800 Retrieving
Narrow Mouth Bottles 15 mL (0.5 oz.) 20-410 89221-584 Case of 2,000 Retrieving
Narrow Mouth Bottles 30 mL (1 oz.) 20-410 414004-154 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 6480 Retrieving
Wide Mouth Bottles 30 mL (1 oz.) 20-410 89221-658 Case of 1,000 Retrieving
Narrow Mouth Bottles 30 mL (1 oz.) 20-410 89221-586 Case of 1,000 Retrieving
Pallet of 6000 Retrieving
Narrow Mouth Bottles 60 mL (2 oz.) 20-410 414004-155 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 6336 Retrieving
Wide Mouth Bottles 60 mL (2 oz.) 20-410 89221-660 Case of 1,000 Retrieving
Pallet of 6000 Retrieving
Narrow Mouth Bottles 60 mL (2 oz.) 20-410 89221-588 Case of 1,000 Retrieving
Narrow Mouth Bottles 125 mL (4.2 oz.) 24-410 414004-156 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 3456 Retrieving
Wide Mouth Bottles 125 mL (4.2 oz.) 24-410 89221-662 Case of 500 Retrieving
Pallet of 4000 Retrieving
Narrow Mouth Bottles 125 mL (4.2 oz.) 24-410 89221-590 Case of 500 Retrieving
Pallet of 4000 Retrieving
Narrow Mouth Bottles 250 mL (8.5 oz.) 24-410 414004-157 Case of 72 Retrieving
Pack of 12 Retrieving
Pallet of 1296 Retrieving
Wide Mouth Bottles 250 mL (8.5 oz.) 24-410 89221-664 Case of 250 Retrieving
Pallet of 1500 Retrieving
Narrow Mouth Bottles 250 mL (8.5 oz.) 24-410 89221-592 Case of 250 Retrieving Purchase Hold
Pallet of 1500 Retrieving Purchase Hold
Narrow Mouth Bottles 500 mL (16.9 oz.) 28-410 414004-158 Case of 48 Retrieving
Pack of 12 Retrieving
Pallet of 1056 Retrieving
Wide Mouth Bottles 500 mL (16.9 oz.) 28-410 89221-666 Case of 125 Retrieving
Pallet of 625 Retrieving
Narrow Mouth Bottles 500 mL (16.9 oz.) 28-410 89221-594 Case of 125 Retrieving
Narrow Mouth Bottles 1 L (33.8 oz.) 38-430 414004-159 Case of 24 Retrieving
Pack of 6 Retrieving
Pallet of 432 Retrieving
Wide Mouth Bottles 1 L (33.8 oz.) 38-430 89221-668 Case of 50 Retrieving
Narrow Mouth Bottles 1 L (33.8 oz.) 38-430 89221-596 Case of 50 Retrieving
Pallet of 400 Retrieving
purchaseHold Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty