Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lacromin™ Holo Recombinant Lactoferrin, InVitria

Supplier: Invitria

Lacromin, recombinant human lactoferrin is an animal component free cell culture media supplement that increases cell growth while protecting cells from apoptosis (programmed cell death).

  • Animal-free
  • Outperforms insulin-like growth factor (IGF)
  • Improved cell growth
  • Cost-effective
  • Replaces IGF
  • Protects cells from apoptosis
  • Well defined and consistent
  • Regulatory friendly
Research has demonstrated that Lacromin often outperforms and is used as a replacement for more expensive Insulin-like Growth Factor (IGF). Lacromin is a simple, yet powerful tool used to optimize media for maximum cell viability and growth. As a recombinant lactoferrin, Lacromin provides animal-free, defined, and consistent performance.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 g 777LAC015-100G 75784-774 Each (100g) Retrieving Restricted
10 g 777LAC015-10G 10847-796 Each (10g) Retrieving
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Conductivity (1% solution) <4 mS
Endotoxin Content <10 EU/mg
Moisture <10%
Protease-free Yes
Protein Type Recombinant
Purity ≥90% (>85% total Protein) ≥90%
Species Human Oryza sativa
Total Iron >1.2 mg/g LF
Total Protein >85%
pH (1% solution) 6.0-8.0
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty