Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lysobac™ Recombinant Lysozyme, InVitria

Supplier: Invitria
Lysobac is a breakthrough for microbial cell lysis and can be used as a preservative in foods and beverages, diagnostic applications, bioprocessing and life science research.

  • Animal-free
  • Higher performance
  • Improved cell lysis
  • Gentle lysis agent
  • Reduced cost
  • Increased consistency

Lysobac is recombinant human lysozyme produced in an animal-free production system. Animal-free production eliminates the risk of adding an undesirable animal component or potential allergen. Lysobac is a more effective preservative than chicken lysozyme and is four times more active per mg for lysing Micrococcus and E. coli when compared to chicken lysozyme.

Lysobac saves bioprocessing time by eliminating cell harvest to provide in-culture protein extraction. In addition, because Lysobac is highly active it adds minimal volume to the final lysate. Lysobac also has the advantage of being consistent across product lots unlike chicken lysozyme, which has a great deal of variability. Lysobac is a gentle lysing agent providing minimal risk of denaturing protein.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 g 777LYS016-10G 10847-774 Each (10g) Retrieving
100 g 777LYS016-100G 10847-776 Each (100g) Retrieving
1 kg 777LYS016-1KG 10847-780 Each (1kg) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Lysobac Lysozyme
Protein/Peptide Type Recombinant
Protease-free Yes
Animal-free Yes
Formulation Powder
Biological Activity >125,000 Units/mg protein
Solubility (1%, A600nm) <2.5
Moisture <7%
Protein Type Recombinant
Total Protein ≥90%
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty