Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Optiferrin™ Recombinant Transferrin, InVitria

Supplier: Invitria
Optiferrin is recombinant human transferrin that is completely animal component free.

  • Animal-free
  • No cell damage
  • Cost-effective
  • Replaces bovine transferrin and plasma derived human transferrin
  • Replaces iron chelators and other iron sources
  • Well defined and consistent

Optiferrin's performance is equivalent to human transferrin in cell culture and offers significant regulatory advantages due to its safety and consistency. Optiferrin is produced in an animal-free system (ExpressTec), which eliminates safety concerns associated with animal or human derived transferrin that can cause contamination with viruses and prions. In addition, Optiferrin is a consistent product with none of the lot-to-lot variability seen in other transferrins and is an excellent animal-free replacement for bovine transferrin and plasma-derived human transferrin.

As a universal iron carrier, Optiferrin also provides better performance than other iron sources and iron chelators. All cells need iron for cell division and transferrin is specifically designed to deliver iron to the cells. Each cell has transferrin receptors that ensure optimal iron delivery based on the actual iron need of the cells. The cells decide how much iron they need, thus eliminating the problem of too much iron causing cell damage. The problem with alternative iron sources and iron chelators is that cells cannot regulate the amount of iron that is delivered and studies have shown that cells can take in 100 times physiological intracellular iron. The uptake of too much iron causes significant cell damage and other iron sources can cause oxidative damage which also causes cell damage; neither of these types of damage can occur when transferrin is used.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 g 777TRF029-1G 10847-770 Each (1g) Retrieving
10 g 777TRF029-10G 10847-778 Each (10g) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Optiferrin Transferrin
Protein/Peptide Type Recombinant
Species Human
Protease-free Yes
Purity >95%
Endotoxin Content ≤1 EU/mg
Formulation Powder, pH (1% solution) 6.0-8.0
Solubility (1%, A600nm) <0.5
Moisture <10%
Protein Type Recombinant
Total Protein >90%
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty