Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cellastim™ Recombinant Human Serum Albumin, InVitria

Supplier: Invitria
Cellastim is recombinant human serum albumin that is completely animal component free.

  • Animal-free
  • Increased performance
  • Cost-effective
  • Applicable to all phases of cell culture

Cellastim has been shown to increase the performance of a wide range of cell types, and is often used as a tool in creating an animal-free cell culture production system. Cellastim helps to facilitate the removal of animal-derived supplements including serum, bovine serum albumin, and human serum albumin without sacrificing performance.

In culture that is already animal-free, Cellastim is an important addition to all phases of the culture process from cell cloning and selection to expansion and commercial manufacturing. As a recombinant albumin, Cellastim provides consistent performance compared to animal-derived supplements. Cellastim can be used in chemically defined media or with plant hydrolysates for optimal performance. With the many benefits Cellastim provides, it is a very cost-effective way to maximize cell culture performance.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 g 777HSA017-100G 10847-716 Each (100g) Retrieving
10 g 777HSA017-10G 10847-718 Each (10g) Retrieving
1 kg 777HSA017-1KG 10847-792 Each (1kg) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Cellastim Albumin
Protein/Peptide Type Recombinant
Species Human
Protease-free Yes
Animal-free Yes
Purity >96%
Endotoxin Content ≤ 1 EU/mg
Formulation Powder
Protein Type Recombinant
Total Protein ≥90%
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty