Would you like to log in again?
ITSE Animal-Free, InVitria
Supplier: Invitria
ITSE Animal-Free (rInsulin-rTransferrin-Selenium-Ethanolamine) is a recombinant insulin and transferrin cell culture media supplement that is completely animal component free.
  • Animal component-free
  • Formulated with selenium and ethanolamine
  • Improved consistency
  • Cost-effective
  • Ready-to-use 100X sterile liquid
  • Sterile
ITSE Animal-Free is formulated with selenium and ethanolamine for improved serum reduced or serum free cell growth.
Insulin, transferrin, selenium, and ethanolamine are typically required for optimal cell growth in serum-free media. Insulin has cell signaling functions and promotes the uptake of glucose and amino acids. Transferrin is a universal source of iron for cells that is a non-toxic carrier of iron and reduces the generation of toxic free radicals and peroxide. Selenium is required for the activity of glutathione peroxidase, thioredoxin reductase, and other antioxidant enzymes. Ethanolamine is a phospholipid precursor that improves the performance of cells in serum-free media and is required for the growth of some cell types.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 mL 777ITS032-10ML 10847-714 Each (10ml) Retrieving
10 x 10 mL 777ITS03210X10 75784-766 Each(10ml) Retrieving Restricted
100 mL 777ITS032100ML 10847-794 Each (100ml) Retrieving
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Conductivity 13.7-16.7 mS/cm
pH 7.7-8.7
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty