Would you like to log in again?
ACTERO™ MediaBox, Foodchek
Supplier: Foodchek
The Actero™ MediaBox™ is the solution for laboratories that require liquid culture media on-demand. Delivering fresh, sterile media in an easy-to-handle, stackable, storage box, Actero™ MediaBox™ provides affordable, ready-to-use liquid culture media in 5 L and 10 L sizes, with a 20 L option available upon request.
While Actero Universal Enrichment media represent fast-acting and established formulations that can be used for standard testing protocols employed in today’s labs, the Patent Pending Actero Elite Enrichment Media represent enhanced formulations that further reduce the required enrichment time to an industry-leading low.
Get even faster time-to-results and accuracy when you combine Actero Enrichment Media with the FoodChek™ MICT Reader and Assay
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Elite Enrichment Media
Description Size Includes Supplier No. VWR Catalog Number Unit Price Quantity
ACTERO™ mTSB 5 L Mediabox™ of Sterile Modified Tryptone Soya Broth for microorganism culture FCM-040 89522-696 Each (5l) Retrieving Restricted
ACTERO™ mTSB 10 L Mediabox™ of Sterile Modified Tryptone Soya Broth for microorganism culture FCM-041 89522-698 Each (10l) Retrieving Restricted
ACTERO™ mTSB 20 L Mediabox™ of Hydrated, Sterile Modified Tryptone Soya Broth for microorganism culture FCM-042 89522-700 Each (20l) Retrieving Restricted
ACTERO™ mUVM 5 L Mediabox™ of Sterile mUVM for selective enrichment of Listeria spp. FCM-043 89522-702 Each (5l) Retrieving Restricted
ACTERO™ mUVM 10 L Mediabox™ of Sterile mUVM for selective enrichment of Listeria spp. FCM-044 89522-704 Each (10l) Retrieving Restricted
ACTERO™ Listeria 5 L Mediabox™ of ACTERO™ Listeria enrichment media for selective enrichment of Listeria spp. FCM-045 89522-706 Each (5l) Retrieving Restricted
ACTERO™ Listeria 10 L Mediabox™ of ACTERO™ Listeria enrichment media for selective enrichment of Listeria spp. FCM-046 89522-708 Each (10l) Retrieving Restricted
ACTERO™ Salmonella 5 L Mediabox™ of ACTERO™ Salmonella enrichment media for selective enrichment of Salmonella spp. FCM-047 89522-710 Each (5l) Retrieving Restricted
ACTERO™ Salmonella 10 L Mediabox™ of ACTERO™ Salmonella enrichment media for selective enrichment of Salmonella spp. FCM-048 89522-712 Each (10l) Retrieving Restricted
Universal Enrichment Media
Description Size Includes Supplier No. VWR Catalog Number Unit Price Quantity
ACTERO™ Sterile Water 5 L Autoclaved, Sterile Water FCM-031 89522-678 Each (5l) Retrieving Restricted
ACTERO™ Sterile Water 10 L Autoclaved, Sterile Water FCM-032 89522-680 Each (10l) Retrieving Restricted
ACTERO™ Phosphate Buffer 5 L Mediabox™ of Sterile Phosphate Buffer for water and waste water analysis (APHA or EPA methods) FCM-033 89522-682 Each (5l) Retrieving Restricted
ACTERO™ Phosphate Buffer 10 L Mediabox™ of Sterile Phosphate Buffer for water and waste water analysis (APHA or EPA methods) FCM-034 89522-684 Each (10l) Retrieving Restricted
ACTERO™ Butterfields 5 L Mediabox™ of Sterile Butterfields for food and dairy products (APHA, FDA, AOAC, or USP methods) FCM-035 89522-686 Each (5l) Retrieving Restricted
ACTERO™ Butterfields 10 L Mediabox™ of Sterile Butterfields for food and dairy products (APHA, FDA, AOAC, or USP methods) FCM-036 89522-688 Each (10l) Retrieving Restricted
ACTERO™ 0.1% Peptone Water 10 L MediaBox™ of 0.1% Peptone Water, Ready-to-Use sample diluent for the preparation of suspensions of microorganisms FCM-113 10146-836 Each (10l) Retrieving Restricted
ACTERO™ 0.1% Peptone Water 5 L MediaBox™ of 0.1% Peptone Water, Ready-to-Use sample diluent for the preparation of suspensions of microorganisms FCM-112 10146-834 Each (5l) Retrieving Restricted
ACTERO™ 0.1% Peptone Water 20 L MediaBox™ of 0.1% Peptone Water, Ready-to-Use sample diluent for the preparation of suspensions of microorganisms FCM-114 10146-838 Each (20l) Retrieving Restricted
ACTERO™ Demi-Fraser Media (Modification of Fraser Broth) 20 L Mediabox™ of Demi-Fraser Media for rapid detection of Listeria with the addition of citrate FCM-129 10159-104 Each (20l) Retrieving Restricted
ACTERO™ Demi-Fraser Media (Modification of Fraser Broth) 5 L Mediabox™ of Demi-Fraser Media for rapid detection of Listeria with the addition of citrate FCM-127 10159-100 Each (5l) Retrieving Restricted
ACTERO™ Demi-Fraser Media (Modification of Fraser Broth) 10 L Mediabox™ of Demi-Fraser Media for rapid detection of Listeria with the addition of citrate FCM-128 10159-102 Each (10l) Retrieving Restricted
ACTERO™ Lactose Broth 20 L MediaBox™ of Lactose Broth for the detection and/or verification of the presence of Salmonella and other coliform organisms FCM-123 10159-106 Each (21l) Retrieving Restricted
ACTERO™ Lactose Broth 5 L MediaBox™ of Lactose Broth for the detection and/or verification of the presence of Salmonella and other coliform organisms FCM-110 10127-232 Each (5l) Retrieving Restricted
ACTERO™ Lactose Broth 10 L MediaBox™ of Lactose Broth for the detection and/or verification of the presence of Salmonella and other coliform organisms FCM-111 10127-234 Each (10l) Retrieving Restricted
ACTERO™ LEB (Basal UVM Formulation) 10 L MediaBox™ of LEB (Basal UVM Formulation) for enrichment of Listeria monocytogenes FCM-119 10146-848 Each (10l) Retrieving Restricted
ACTERO™ LEB (Basal UVM Formulation) 20 L MediaBox™ of LEB (Basal UVM Formulation) for enrichment of Listeria monocytogenes FCM-120 10146-850 Each (20l) Retrieving Restricted
ACTERO™ LEB (Basal UVM Formulation) 5 L MediaBox™ of LEB (Basal UVM Formulation) for enrichment of Listeria monocytogenes FCM-118 10146-846 Each (5l) Retrieving Restricted
ACTERO™ Modified Tryptic Soy Broth with Cassamino 5 L MediaBox™ of Modified Tryptic Soy Broth with Cassamino for microorganism culture FCM-240 10500-814 Each (5l) Retrieving Restricted
ACTERO™ Tryptic Soy Broth 5 L MediaBox™ of Tryptic Soy Broth for the enrichment and cultivation of aerobic microorganisms FCM-236 10500-806 Each (5l) Retrieving Restricted
ACTERO™ Tryptic Soy Broth 10 L MediaBox™ of Tryptic Soy Broth for the enrichment and cultivation of aerobic microorganisms FCM-237 10500-808 Each (10l) Retrieving Restricted
ACTERO™ Universal Pre-enrichment Broth 20 L Mediabox™ of Universal Pre-enrichment Broth, Non-selective medium for recovery of Salmonella and Listeria from food FCM-207 10159-098 Each (20l) Retrieving Restricted
ACTERO™ Universal Pre-enrichment Broth 5 L Mediabox™ of Universal Pre-enrichment Broth, Non-selective medium for recovery of Salmonella and Listeria from food FCM-205 10159-094 Each (5l) Retrieving Restricted
ACTERO™ Universal Pre-enrichment Broth 10 L Mediabox™ of Universal Pre-enrichment Broth, Non-selective medium for recovery of Salmonella and Listeria from food FCM-206 10159-096 Each (10l) Retrieving Restricted
ACTERO™ UVM1 Complete Media 5 L MediaBox™ of UVM1 Complete Media for the initial enrichment of Listeria monocytogenes FCM-115 10146-840 Each (5l) Retrieving Restricted
ACTERO™ UVM1 Complete Media 20 L MediaBox™ of UVM1 Complete Media for the initial enrichment of Listeria monocytogenes FCM-117 10146-844 Each (20l) Retrieving Restricted
ACTERO™ UVM1 Complete Media 10 L MediaBox™ of UVM1 Complete Media for the initial enrichment of Listeria monocytogenes FCM-116 10146-842 Each (10l) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Female Adapter for 6 mm I.D. Tubing, Sterile, Autoclavable Adapter Only, Tubing Sold Separately FCLM-031 10127-246 Each Retrieving Restricted
Female Adapter for 8 mm I.D. Tubing, Sterile, Autoclavable Adapter Only, Tubing Sold Separately FCLM-028 10127-240 Each Retrieving Restricted
Silicone Tubing Set, Nonsterile, Autoclavable, 6 mm I.D. Five feet of tubing with female adapter, capped FCLM-030 10127-244 Each Retrieving Restricted
Silicone Tubing Set, Nonsterile, Autoclavable, 8 mm I.D. Five feet of tubing with female adapter, capped FCLM-027 10127-238 Each Retrieving Restricted
Silicone Tubing Set, Sterile, Autoclavable, 6 mm I.D. Five feet of tubing with female adapter, capped in a sealed bag FCLM-029 10127-242 Each Retrieving Restricted
Silicone Tubing Set, Sterile, Autoclavable, 8 mm I.D. Five feet of tubing with female adapter, capped in a sealed bag FCLM-026 10127-236 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty